PloS one
Cavin-3 knockout mice show that cavin-3 is not essential for caveolae formation, for maintenance of body composition, or for glucose tolerance.
PMID 25036884
Abstract
The cavins are a family of proteins associated with caveolae, cavin-1, -2 and -3 being widely expressed while cavin-4 is restricted to striated muscle. Deletion of cavin-1 results in phenotypes including metabolic changes consistent with adipocyte dysfunction, and caveolae are completely absent. Deletion of cavin-2 causes tissue-specific loss of caveolae. The consequences of cavin-3 deletion are less clear, as there are divergent data on the abundance of caveolae in cavin-3 null mice. Here we examine the consequences of cavin-3 deficiency in vivo by making cavin-3 knockout mice. We find that loss of cavin-3 has minimal or no effects on the levels of other caveolar proteins, does not appear to play a major role in formation of protein complexes important for caveolar morphogenesis, and has no significant effect on caveolae abundance. Cavin-3 null mice have the same body weight and fat mass as wild type animals at ages 8 through 30 weeks on both normal chow and high fat diets. Likewise, the two mouse strains exhibit identical glucose tolerance tests on both diets. Microarray analysis from adipose tissue shows that the changes in mRNA expression between cavin-3 null and wild type mouse are minimal. We conclude that cavin-3 is not absolutely required for making caveolae, and suggest that the mechanistic link between cavin-3 and metabolic regulation remains uncertain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2900061
Anti-CAV1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900062
Anti-CAV1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV09019
Anti-CAV1 antibody produced in rabbit, purified antibody, Protein A purified
SAB2900063
Anti-CAV1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100344
Anti-CAV1 antibody produced in rabbit, affinity isolated antibody
C4490
Anti-Caveolin-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
C3237
Anti-Caveolin-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1408474
Anti-PTRF antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV36965
Anti-PTRF antibody produced in rabbit, IgG fraction of antiserum
HPA049838
Anti-PTRF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410563
Anti-PTRF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2502166
Anti-TFRC (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA028598
Anti-TFRC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200398
Anti-TFRC antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200216
Monoclonal Anti-Caveolin-1 antibody produced in mouse, clone CAV1, tissue culture supernatant
SAB4700515 Monoclonal Anti-CD71 antibody produced in mouse, clone MEM-75, purified immunoglobulin, buffered aqueous solution
SAB4700520 Monoclonal Anti-CD71 antibody produced in mouse, clone MEM-189, purified immunoglobulin, buffered aqueous solution
WH0284119M2
Monoclonal Anti-PTRF antibody produced in mouse, clone 1F7, purified immunoglobulin, buffered aqueous solution
SAB1402945
Monoclonal Anti-TFRC antibody produced in mouse, clone 1E6, purified immunoglobulin, buffered aqueous solution