EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Heterologous production, purification, and immunodetection of Magnaporthe oryzae avirulence protein AVR-Pia.
PMID 25036967
Abstract
The avirulence gene AVR-Pia of Magnaporthe oryzae, which induces a hypersensitive reaction in rice cultivars containing the resistance gene Pia, was expressed in Escherichia coli. AVR-Pia protein was collected as inclusion bodies, denatured, and refolded. Finally, recombinant AVR-Pia (rAVR-Pia) protein was purified by column chromatography. Infiltration of rAVR-Pia triggered cell browning in the leaves of rice cultivar Aichiasahi (Pia), with accumulation of H2O2 and induction of PR1a expression in rice. On the other hand, these reactions were not observed in Shin-2 (pia) leaves after the same treatment. This observation indicated that rAVR-Pia had the function of an avirulence protein. rAVR-Pia was used for immunization of a rabbit, and anti-AVR-Pia antiserum was prepared. The specificity of this antibody was appraised by detecting native AVR-Pia in the inoculated leaf sheath extract using Western blotting in combination with immunoprecipitation. Native AVR-Pia was successfully detected, and its molecular weight was estimated to be 7.4 kDa, indicating signal peptide cleavage. Additionally, secreted native AVR-Pia was quantified as 3.7 ng/g rice sheath.