EMAIL THIS PAGE TO A FRIEND
The Annals of thoracic surgery
Antimicrobial peptides are highly abundant and active in postoperative pleural drainage fluids.
PMID 25038012
Abstract
The human lung is considered a nonsterile organ, and surgical interventions therefore take place in a more or less contaminated operating field. Nevertheless, infectious complications of the pleural cavity are low after major lung resections. Antimicrobial peptides (AMPs) are part of the innate immunity and display a broad capacity to kill pathogens. We hypothesized that the pleural space must have a high natural antimicrobial barrier and that AMPs might effectively protect the pleural cavity. Pleural effusions were collected after lung operations. Antimicrobial activity of the fluids against gram-positive and gram-negative pathogens was analyzed by microdilution assays. AMPs were determined by enzyme-linked immunosorbent assay (ELISA), polymerase chain reaction (PCR), and immunohistochemical analysis. The impact of proinflammatory triggers on AMP release from pleural mesothelial cells was evaluated. Antimicrobial activity assays revealed high bactericidal properties of postoperative pleural drainage fluids. They effectively killed gram-negative pathogens (Escherichia coli, Pseudomonas aeruginosa) as well as gram-positive pathogens (Staphylococcus aureus, Streptococcus pneumoniae, and Streptococcus pyogenes). A variety of AMPs was detected at constantly high concentrations in the pleural fluids. They mainly derived from leukocytes and pleural epithelium. Although proinflammatory cytokine levels were elevated in the postoperative pleural fluids, AMP expression could not be augmented by Toll-like receptor (TLR) triggering or by the proinflammatory cytokines interleukin (IL)-1β and tumor necrosis factor (TNF)α. We provide the first evidence of a high abundance of AMPs in postoperative pleural fluids. Our findings might explain the broad protection against infectious complications of the pleural space after major lung operations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW23003 Anti-CD19 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405577
Anti-CD8A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA037756
Anti-CD8A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000930M1
Monoclonal Anti-CD19 antibody produced in mouse, clone 1G3, purified immunoglobulin, buffered aqueous solution
SAB1403645
Monoclonal Anti-CD19 antibody produced in mouse, clone 1C9, purified immunoglobulin, buffered aqueous solution
SAB4700108 Monoclonal Anti-CD19 antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB4700109 Monoclonal Anti-CD19 antibody produced in mouse, clone LT19, purified immunoglobulin, buffered aqueous solution
SAB4700059
Monoclonal Anti-CD4 , (N-terminal) antibody produced in mouse, clone MEM-241, purified immunoglobulin, buffered aqueous solution
SAB4700054 Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-16, purified immunoglobulin, buffered aqueous solution
SAB4700065
Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-115, purified immunoglobulin, buffered aqueous solution
SAB4700733 Monoclonal Anti-CD4 antibody produced in mouse, clone OX-35, purified immunoglobulin, buffered aqueous solution
SAB4100489
Monoclonal Anti-CD4 antibody produced in mouse, clone CD4 -A409, culture supernatant
AMAB90754
Monoclonal Anti-CD4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0395, purified immunoglobulin, buffered aqueous glycerol solution
C7423
Monoclonal Anti-CD8 antibody produced in mouse, clone UCHT-4, purified immunoglobulin, buffered aqueous solution
SAB4700084 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-31, purified immunoglobulin, buffered aqueous solution
SAB4700089 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-87, purified immunoglobulin, buffered aqueous solution
SAB1402142
Monoclonal Anti-CD8A antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution