EMAIL THIS PAGE TO A FRIEND
Food chemistry
A study of multi-ligand beta-lactoglobulin complex formation.
PMID 25038674
Abstract
Beta-lactoglobulin (β-LG), the principal whey protein, possesses multiple sites for binding ligands. Most studies of β-LG-ligand interactions have focused on the formation and dissociation of protein complexes with single ligands, such as α-tocopherol, resveratrol or folic acid. In this study, the possibility of a plurality of bioactive compounds binding simultaneously to β-LG was analysed using protein intrinsic fluorescence quenching. It was found that β-LG could bind two or three ligands simultaneously, although the sequence in which the ligands were added affected binding affinity. The impact of binding to β-LG on physicochemical properties of these three ligands is discussed in view of fluorescence spectroscopy and high performance liquid chromatography results. The data obtained in this study suggest the feasibility of developing β-LG-based carriers of a plurality of active compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7876
Folic acid, ≥97%
C19H19N7O6
1286005
Folic acid, United States Pharmacopeia (USP) Reference Standard
C19H19N7O6
F8758
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
C19H19N7O6
F8798
Folic acid, meets USP testing specifications
C19H19N7O6
01769
Folic acid, tested according to Ph.Eur.
C19H19N7O6
F0300000
Folic acid, European Pharmacopoeia (EP) Reference Standard
C19H19N7O6
PHR1035
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C19H19N7O6
47866
Folic acid, analytical standard
C19H19N7O6
38364
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
HNa2O4P
R5010
Resveratrol, ≥99% (HPLC)
C14H12O3
76511
Resveratrol, certified reference material, TraceCERT®
C14H12O3
34092
Resveratrol, analytical standard
C14H12O3
Y0001194
Resveratrol, European Pharmacopoeia (EP) Reference Standard
C14H12O3
S7907
Sodium phosphate dibasic, BioXtra, ≥99.0%
HNa2O4P
S3264
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
HNa2O4P
S5136
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
HNa2O4P
255793
Sodium phosphate dibasic, 99.95% trace metals basis
HNa2O4P
S9763
Sodium phosphate dibasic, ACS reagent, ≥99.0%
HNa2O4P
S0876
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
HNa2O4P
71636
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
HNa2O4P
71639
Sodium phosphate dibasic, BioUltra, anhydrous, ≥99.5% (T)
HNa2O4P
71640
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
HNa2O4P
04276
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
HNa2O4P
71642
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
HNa2O4P
W239901
Sodium phosphate dibasic, 99%, FG
HNa2O4P
94046
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
HNa2O4P
RDD007
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
H2NaO4P
S8282
Sodium phosphate monobasic, BioXtra, ≥99.0%
H2NaO4P
S3139
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
H2NaO4P
S5011
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
H2NaO4P