EMAIL THIS PAGE TO A FRIEND
Journal of molecular neuroscience : MN
Protein profiling reveals antioxidant and signaling activities of NAP (Davunetide) in rodent hippocampus exposed to hypobaric hypoxia.
PMID 25038875
Abstract
NAP (davunetide) is a clinical octapeptide and reportedly possesses neuroprotective, neurotrophic and cognitive protective properties. The information for NAP-mediated neuroproteome changes and associated signaling pathways during hypoxia will help in drug development programmes across the world. In the present study, we have evaluated the antioxidant activities of NAP in rat hippocampus exposed to hypobaric hypoxia (25,000 ft, 282 mm Hg) for 3, 6 and 12 h respectively. Using 2D-gel electrophoresis (2D-GE) with matrix-assisted laser desorption ionization time of flight (MALDI-TOF/TOF) mass spectrometry, we have identified altered expression of 80 proteins in NAP-supplemented hippocampus after hypoxia. Pathway analysis revealed that NAP supplementation significantly regulated oxidative stress response, oxidoreductase activity and cellular response to stress pathways during hypoxia. Additionally, NAP supplementation also regulated energy production pathways along with AMP-activated protein kinase (AMPK) signaling and signaling by Rho family GTPases pathways. We observed higher expression of antioxidant Sod1, Eno1, Prdx2 and Prdx5 proteins that were subsequently validated by Western blotting. A higher level of Prdx2 was also observed by immunohistochemistry in NAP-supplemented hippocampus during hypoxia. In corroboration, we are able to detect significant lower level of protein carbonyls in NAP-supplemented hypoxic hippocampus suggesting amelioration of oxidant molecules by NAP supplementation. These results emphasize the antioxidant and signaling properties of NAP in rodent hippocampus during hypobaric hypoxia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2101878
Anti-PRDX2 antibody produced in rabbit, affinity isolated antibody