EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Increased expression of stress inducible protein 1 in glioma-associated microglia/macrophages.
PMID 25042352
Abstract
Factors released by glioma-associated microglia/macrophages (GAMs) play an important role in the growth and infiltration of tumors. We have previously demonstrated that the co-chaperone stress-inducible protein 1 (STI1) secreted by microglia promotes proliferation and migration of human glioblastoma (GBM) cell lines in vitro. In the present study, in order to investigate the role of STI1 in a physiological context, we used a glioma model to evaluate STI1 expression in vivo. Here, we demonstrate that STI1 expression in both the tumor and in the infiltrating GAMs and lymphocytes significantly increased with tumor progression. Interestingly, high expression of STI1 was observed in macrophages and lymphocytes that infiltrated brain tumors, whereas STI1 expression in the circulating blood monocytes and lymphocytes remained unchanged. Our results correlate, for the first time, the expression of STI1 and glioma progression, and suggest that STI1 expression in GAMs and infiltrating lymphocytes is modulated by the brain tumor microenvironment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV46164
Anti-STIP1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV46165
Anti-STIP1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
GW22824 Anti-STIP1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407079
Anti-STIP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA044062
Anti-STIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039291
Anti-STIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0084525M1
Monoclonal Anti-HOP antibody produced in mouse, clone 3D6, purified immunoglobulin, buffered aqueous solution
WH0010963M1
Monoclonal Anti-STIP1 antibody produced in mouse, clone 4B6, purified immunoglobulin, buffered aqueous solution
SAB1408802
Monoclonal Anti-STIP1 antibody produced in mouse, clone 2E1, purified immunoglobulin, buffered aqueous solution
SAB1408803
Monoclonal Anti-STIP1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
SAB1408804
Monoclonal Anti-STIP1 antibody produced in mouse, clone 1C6, purified immunoglobulin, buffered aqueous solution
SAB1408805
Monoclonal Anti-STIP1 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution