EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
A polythiophene derivative with superior properties for practical application in polymer solar cells.
PMID 25044098
Abstract
A polythiophene derivative called PDCBT, which has a backbone of thiophene units and just carboxylate functional groups to modulate its properties, exhibits properties superior to those of poly(3-hexylthiophene), the classic polythiophene derivative, when used as an electron donor in polymer solar cells (PSCs). The best device, based on PDCBT/PC71BM (1:1), develops a good power conversion efficiency of 7.2%.