EMAIL THIS PAGE TO A FRIEND
Cell biology international
Tribbles homolog 3 expression in spermatogonial stem cells of rat testes.
PMID 25044179
Abstract
Tribbles are a family of signal-regulating proteins shown to coordinate the action and the suppression of different pathways. Tribbles homolog 3 (Trib3), the best-studied member of the mammalian tribble family, has a key function in determining cell fate when responding to environmental challenges. Trib3 effects are also modulated by its direct interaction with other signaling molecules. We found that Trib3 is highly expressed in the early development of rat testis, at just the time when the gonocytes resume proliferation to give rise to A spermatogonia. Immunofluorescence staining of cross-sections of rat testis and cultured spermatogonial stem cells (SSCs) also confirmed that Trib3 is expressed in rat SSCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32790
1,4-Diaminobutane, puriss., ≥99.0% (GC)
C4H12N2
32791
1,4-Diaminobutane, purum, ≥98.0% (GC)
C4H12N2
HPA043536
Anti-FZR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106927
Anti-FZR1 antibody produced in rabbit, affinity isolated antibody
SAB2105778
Anti-FZR1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1407812
Anti-TRIB3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA015272
Anti-TRIB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411157
Anti-TRIB3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
L2376
Linolenic acid, ≥99%
C18H30O2
62170
Linolenic acid, ~70% (GC)
C18H30O2
62160
Linolenic acid, analytical standard
C18H30O2
SAB1402580
Monoclonal Anti-FZR1 antibody produced in mouse, clone 4C4, purified immunoglobulin, buffered aqueous solution
SAB1405091
Monoclonal Anti-FZR1, (N-terminal) antibody produced in mouse, clone 3E12, purified immunoglobulin, buffered aqueous solution
WH0057761M3
Monoclonal Anti-TRIB3 antibody produced in mouse, clone 1H2, purified immunoglobulin, buffered aqueous solution
SAB1405226
Monoclonal Anti-TRIB3 antibody produced in mouse, clone 2F7, purified immunoglobulin, buffered aqueous solution
SAB1409582
Monoclonal Anti-TRIB3 antibody produced in mouse, clone 3D7, purified immunoglobulin, buffered aqueous solution
P9417
Palmitoleic acid, ≥98.5% (GC), liquid
C16H30O2
76169
Palmitoleic acid, analytical standard
C16H30O2
51799
Putrescine, analytical standard
C4H12N2