EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Chlorine-free pyrotechnics: copper(I) iodide as a "green" blue-light emitter.
PMID 25044436
Abstract
The generation of blue-light-emitting pyrotechnic formulations without the use of chlorine-containing compounds is reported. Suitable blue-light emission has been achieved through the generation of molecular emitting copper(I) iodide. The most optimal copper(I) iodide based blue-light-emitting formulation was found to have performances exceeding those of chlorine-containing compositions, and was found to be insensitive to various ignition stimuli.