EMAIL THIS PAGE TO A FRIEND
BioMed research international
Multipotent neural crest stem cell-like cells from rat vibrissa dermal papilla induce neuronal differentiation of PC12 cells.
PMID 25045659
Abstract
Bone marrow mesenchymal stem cells (BMSCs) transplants have been approved for treating central nervous system (CNS) injuries and diseases; however, their clinical applications are limited. Here, we model the therapeutic potential of dermal papilla cells (DPCs) in vitro. DPCs were isolated from rat vibrissae and characterized by immunocytofluorescence, RT-PCR, and multidifferentiation assays. We examined whether these cells could secrete neurotrophic factors (NTFs) by using cocultures of rat pheochromocytoma cells (PC12) with conditioned medium and ELISA assay. DPCs expressed Sox10, P75, Nestin, Sox9, and differentiated into adipocytes, osteoblasts, smooth muscle cells, and neurons under specific inducing conditions. The DPC-conditioned medium (DPC-CM) induced neuronal differentiation of PC12 cells and promoted neurite outgrowth. Results of ELISA assay showed that compared to BMSCs, DPCs secreted more brain-derived neurotrophic factor (BDNF) and glial cell line-derived neurotrophic factor (GDNF). Moreover, we observed that, compared with the total DPC population, sphere-forming DPCs expressed higher levels of Nestin and P75 and secreted greater amounts of GDNF. The DPCs from craniofacial hair follicle papilla may be a new and promising source for treating CNS injuries and diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7018
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
C10H14N4O2
I5879
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
C10H14N4O2
05-1424
Anti-Endoglin Antibody, clone P3D1, 100 µg, Anti-Endoglin Antibody, clone P3D1 is an antibody against Endoglin for use in WB, IH, IC, IP, FC, ELISA.
AB1554
Anti-Nerve Growth Factor Receptor Antibody, p75, 100 µL, Anti-Nerve Growth Factor Receptor Antibody, p75 is an antibody against Nerve Growth Factor Receptor for use in IC, IH(P), and IP.
MAB5262
Anti-Neurofilament 200 kDa Antibody, clone RT97, 50 µg, Anti-Neurofilament 200 kDa Antibody, clone RT97 is an antibody against Neurofilament 200 kDa for use in IH, IH(P) & WB.
AB5535
Anti-Sox9 Antibody, 100 µg, Anti-Sox9 Antibody is a well characterized affinity purified Rabbit Polyclonal Antibody that reliably detects Transcription Factor Sox-9. This highly published antibody has been validated in IHC & WB.
MAB1406
Anti-Thy-1.1 Antibody, clone OX-7, 100 µg, Anti-Thy-1.1 Antibody, clone OX-7 detects level of Thy-1.1 & has been published & validated for use in FC, IC, IH.
D9184
Dexamethasone, meets USP testing specifications
C22H29FO5
D0700000
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
1176007
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
C22H29FO5
46165
Dexamethasone, VETRANAL, analytical standard
C22H29FO5
31375
Dexamethasone, tested according to Ph.Eur.
C22H29FO5
PHR1526
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
C22H29FO5
Y0001593
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
Y0001177
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
03609
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
C10H16N2O8
03620
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
C10H16N2O8
03690
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
C10H16N2O8
F6886
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
C22H34O7
F3917
Forskolin, For use in molecular biology applications
C22H34O7
93049
Forskolin, analytical standard
C22H34O7