EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Unprecedented role of hydronaphthoquinone tautomers in biosynthesis.
PMID 25047689
Abstract
Quinones and hydroquinones are among the most common cellular cofactors, redox mediators, and natural products. Here, we report on the reduction of 2-hydroxynaphthoquinones to the stable 1,4-diketo tautomeric form of hydronaphthoquinones and their further reduction by fungal tetrahydroxynaphthalene reductase. The very high diastereomeric and enantiomeric excess, together with the high yield of cis-3,4-dihydroxy-1-tetralone, exclude an intermediary hydronaphthoquinone. Labeling experiments with NADPH and NADPD corroborated the formation of an unexpected 1,4-diketo tautomeric form of 2-hydroxyhydronaphthoquinone as a stable intermediate. Similar 1,4-diketo tautomers of hydronaphthoquinones were established as products of the NADPH-dependent enzymatic reduction of other 1,4-naphthoquinones, and as substrates for different members of the superfamily of short-chain dehydrogenases. We propose an essential role of hydroquinone diketo tautomers in biosynthesis and detoxification processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51455
Vitamin K3 2,3-epoxide, ≥98.5% (HPLC)
C11H8O3