EMAIL THIS PAGE TO A FRIEND
Chemosphere
Particulate and colloidal silver in sewage effluent and sludge discharged from British wastewater treatment plants.
PMID 25048887
Abstract
Differential filtration was used to measure silver (>2 nm) entering and leaving nine sewage treatment plants (STPs). The mean concentration of colloidal (2-450 nm) silver, which includes nanosilver, was found to be 12 ng L(-1) in the influent and 6 ng L(-1) in the effluent. For particulate silver (>450 nm) the mean values were 3.3 μg L(-1) for influent and 0.08 μg L(-1) for effluent. Thus, removal was around 50% and 98% for colloidal and particulate silver respectively. There was no significant difference in performance between the different types of STP investigated (three examples each of activated sludge, biological filter and biological filter with tertiary treatment located across England, UK). In addition, treated sewage sludge samples (biosolids) were taken from several STPs to measure the total silver likely to be discharged to soils. Total silver was 3-14 mg kg(-1) DW in the sludge (median 3.6), which if the sludge were added at the recommended rate to soil, would add 11 μg kg(-1) yr(-1) to the top 20 cm soil layer. Predicted concentrations using the LF2000-WQX model for all the rivers of England and Wales for nanosilver were typically in the 0-1 ng L(-1) range but levels up to 4 ng L(-1) are possible in a high discharge and low flow scenario. Predicted concentrations for the total particulate forms were mostly below 50 ng L(-1) except for a high discharge and low flow scenario where concentrations could reach 135 ng L(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF30673661
Silver, insulated wire, 10m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Ag
GF00665861
Silver, insulated wire, 10m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Ag
GF75333517
Silver, insulated wire, 10m, conductor diameter 0.05mm, insulation thickness 0.0075mm, polyester insulation, 99.99%
Ag
GF81067394
Silver, insulated wire, 10m, conductor diameter 0.075mm, insulation thickness 0.010mm, nylon insulation, 99.99%
Ag
GF97012773
Silver, insulated wire, 10m, conductor diameter 0.075mm, insulation thickness 0.010mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Ag
GF19750990
Silver, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Ag
GF48834311
Silver, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyimide insulation, 99.9%
Ag
GF52232655
Silver, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.015mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Ag
GF30804487
Silver, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Ag
GF70247268
Silver, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.015mm, epoxy insulation, 99.99%
Ag
GF08856658
Silver, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.023mm, polyester insulation, 99.99%
Ag
GF52562493
Silver, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.024mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Ag
GF60364199
Silver, insulated wire, 1m, conductor diameter 0.075mm, insulation thickness 0.010mm, nylon insulation, 99.99%
Ag
GF09632921
Silver, insulated wire, 1m, conductor diameter 0.075mm, insulation thickness 0.010mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Ag
GF55363855
Silver, insulated wire, 1m, conductor diameter 0.25mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Ag
GF34208388
Silver, insulated wire, 20m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Ag
GF98265066
Silver, insulated wire, 20m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Ag
GF16689347
Silver, insulated wire, 20m, conductor diameter 0.05mm, insulation thickness 0.0075mm, polyester insulation, 99.99%
Ag
GF09219797
Silver, insulated wire, 20m, conductor diameter 0.075mm, insulation thickness 0.010mm, nylon insulation, 99.99%
Ag
GF26596312
Silver, insulated wire, 20m, conductor diameter 0.075mm, insulation thickness 0.010mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Ag
GF68874755
Silver, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Ag
GF21307924
Silver, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyimide insulation, 99.9%
Ag
GF29081572
Silver, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.015mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Ag
GF77384649
Silver, insulated wire, 20m, conductor diameter 0.25mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Ag
GF80907532
Silver, insulated wire, 20m, conductor diameter 0.25mm, insulation thickness 0.015mm, epoxy insulation, 99.99%
Ag
GF07081635
Silver, insulated wire, 20m, conductor diameter 0.25mm, insulation thickness 0.023mm, polyester insulation, 99.99%
Ag
GF70311596
Silver, insulated wire, 20m, conductor diameter 0.25mm, insulation thickness 0.024mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Ag
GF24539564
Silver, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Ag
GF70018129
Silver, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Ag
GF07194797
Silver, microfoil, disks, 10mm, thinness 1.0μm, specific density 1098μg/cm2, permanent mylar 3.5μm support, 99.95%
Ag