EMAIL THIS PAGE TO A FRIEND
Chemosphere
Comparing three approaches in extending biotic ligand models to predict the toxicity of binary metal mixtures (Cu-Ni, Cu-Zn and Cu-Ag) to lettuce (Lactuca sativa L.).
PMID 25048917
Abstract
Metals are always found in the environment as mixtures rather than as solitary elements. However, effect models such as biotic ligand models (BLMs) are usually derived for toxicity prediction of single metals. Our study aimed at predicting mixture toxicity of Cu-Ni, Cu-Zn and Cu-Ag combinations to lettuce (Lactucasativa L.) by combining BLMs with three toxicity indexes: the toxic unit, the overall amounts of metal ions bound to the biotic ligands and the toxic equivalency factor. The accumulation of metal ions at the biotic ligands was used to determine the toxic potency of metals alone or in combination. On the basis of parameters derived from toxicity assessment of individual metals, these three extended BLMs appeared to be all acceptable (p<0.0001) in assessing toxicity of diverse metal mixtures. The BLM-based approaches integrated competition between metal ions in assessing mixture toxicity and showed different predictive ability for each metal combination. The outcome of modeling suggested that the combined toxicity depends on the specific components of the metal mixtures. The best developed models assist in identifying the type of underlying toxic mechanisms of diverse metal mixtures in terrestrial plants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF71667366
Copper, wire reel, 1000m, diameter 0.04mm, annealed, 99.9%
Cu
GF21709650
Copper, wire reel, 1000m, diameter 0.04mm, hard, 99.9%
Cu
GF99677165
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Cu
GF08494577
Copper, wire reel, 1000m, diameter 0.075mm, annealed, 99.9%
Cu
GF66965956
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Cu
GF22319872
Copper, wire reel, 1000m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu
GF27350287
Copper, wire reel, 1000m, diameter 0.1mm, annealed, 99.9%
Cu
GF63655780
Copper, wire reel, 1000m, diameter 0.25mm, annealed, 99.9%
Cu
GF47171033
Copper, wire reel, 100m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF18312791
Copper, wire reel, 100m, diameter 0.02mm, hard, 99.99+%
Cu
GF89712333
Copper, wire reel, 100m, diameter 0.5mm, as drawn, 99.98+%
Cu
GF93417105
Copper, wire reel, 100m, diameter 1.0mm, as drawn, 99.98+%
Cu
GF64298829
Copper, wire reel, 100m, diameter 0.02mm, annealed, 99.99%
Cu
GF86528155
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Cu
GF39045090
Copper, wire reel, 100m, diameter 0.075mm, annealed, 99.9%
Cu
GF71409089
Copper, wire reel, 100m, diameter 0.125mm, annealed, 99.9%
Cu
GF42860651
Copper, wire reel, 100m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu
GF52128008
Copper, wire reel, 100m, diameter 0.1mm, annealed, 99.9%
Cu
GF69188079
Copper, wire reel, 100m, diameter 0.25mm, annealed, 99.9%
Cu
GF98066054
Copper, wire reel, 100m, diameter 0.5mm, annealed, 99.9%
Cu
GF81594399
Copper, wire reel, 100m, diameter 0.5mm, annealed, silver plated, 99.9%
Cu
GF82471620
Copper, wire reel, 100m, diameter 1.0mm, annealed, 99.9%
Cu
GF83100619
Copper, wire reel, 10m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF80026770
Copper, wire reel, 10m, diameter 0.02mm, hard, 99.99+%
Cu
GF74431324
Copper, wire reel, 10m, diameter 0.05mm, as drawn, 99.99+%
Cu
GF81301609
Copper, wire reel, 10m, diameter 0.75mm, as drawn, 99.98+%
Cu
GF86675439
Copper, wire reel, 10m, diameter 0.01mm, hard, 99.99%
Cu
GF77356109
Copper, wire reel, 10m, diameter 0.02mm, annealed, 99.99%
Cu
GF56973257
Copper, wire reel, 2000m, diameter 0.25mm, annealed, 99.9%
Cu