EMAIL THIS PAGE TO A FRIEND
Chemosphere
Assessments of chromium (and other metals) in vegetables and potential bio-accumulations in humans living in areas affected by tannery wastes.
PMID 25048934
Abstract
Chromium (Cr) commonly enters the food chain through uptake by vegetables. However, accurate prediction of plant uptake of Cr (and other metals) still remains a challenge. In this study, we evaluated 5 indices of availability for Cr (and other metals) to identify reliable predictors of metal transfer from soils to garlic, onion, bokchoy, radish and celery grown in soils impacted by tannery wastes. The potential bio-accumulation of Cr in humans was calculated from the Cr content of vegetable predicted by the best bio-availability index, amounts of vegetable consumed and recommended daily doses for Cr. Our results show that soil total Cr is the best predictor of Cr transfer from soils to onion (Cr in onion=8.51+0.005 Total Cr) while Cr extractable by Synthetic Precipitation Leaching Procedure at pH 5 correlates very well with Cr uptake by bokchoy (Cr bokchoy=5.86+7.32 SPLP-5 Cr) and garlic (Cr garlic=7.63+2.36 SPLP-5 Cr). The uptake of Cr by radish and celery could not be reliably estimated by any of the 5 indices of availability tested in this study. Potential bio-accumulation of Cr in humans (BA-Cr) increases from soils with low Cr (BA-Cr=11.5) to soil with high total Cr (BA-Cr=31.3). Due to numerous soil factors affecting the behavior of Cr in soils and the physiological differences among vegetables, we suggest that the prediction of the transfer of Cr (and other metals) from soils to plants should be specific to site, metal and vegetable. Potential bio-accumulation of Cr in humans can be derived from a transfer function of Cr from soils to plants and the human consumption of vegetables.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF22618739
Chromium, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF07622759
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Cr
GF35390651
Chromium, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF46122645
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF31792956
Chromium, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF91240994
Chromium, foil, not light tested, 25x25mm, thickness 0.075mm, permanent polyester support, 99.99+%
Cr
GF98190861
Chromium, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF76896400
Chromium, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF64373766
Chromium, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF34299748
Chromium, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF31161754
Chromium, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr