EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
High glucose induces Rho/ROCK-dependent visfatin and type I procollagen expression in rat primary cardiac fibroblasts.
PMID 25050741
Abstract
Myocardial fibrosis and excessive proliferation of cardiac fibroblasts (CFs) contribute to diabetic cardiomyopathy (DCM). However, the underlying mechanism is still not completely clear. The aim of this study was to investigate the relationship between high-glucose treatment and the expression of visfatin and type I procollagen in rat CFs, and examine the regulatory effects of high‑glucose treatment on the Rho/ROCK signaling pathway. CFs from newborn Sprague Dawley rats were treated with high concentrations of glucose (10, 30 and 50 mmol/l D-glucose), a baseline concentration of glucose (5.5 mmol/l) as a control, and mannitol (5.5 mmol/l D-glucose + 44.5 mmol/l mannitol) as an osmotic control. CFs were also treated with 30 mmol/l D-glucose for 6, 12, 24 and 48 h. The proliferation of CFs was determined by the MTT assay. The mRNA and protein expression of visfatin and type I procollagen were quantified by RT-qPCR and western blot analysis, respectively. Cardiac fibroblast proliferation reached a peak at 30 mmol/l D-glucose, and visfatin and type I procollagen expression were significantly increased upon treatment with high concentrations of glucose (10 and 30 mmol/l) compared to baseline glucose treatment. Treatment with 30 mmol/l D-glucose time-dependently promoted cardiac fibroblast proliferation. The mRNA and protein expression of visfatin and type I procollagen were significantly increased compared to the control at 24 h after 30 mmol/l D-glucose treatment. Y27632, a Rho-associated protein kinase (ROCK) inhibitor, significantly decreased the mRNA and protein levels of visfatin and type I procollagen, induced by 30 mmol/l D-glucose (all P<0.05). In conclusion, a high level of glucose promotes cardiac fibroblast proliferation, and induces visfatin and type I procollagen expression in CFs, at least partially via the Rho/ROCK signaling pathway. These results may be helpful in developing an appropriate therapeutic strategy for DCM.