PloS one
Histone acetyltransferase p300 mediates histone acetylation of PS1 and BACE1 in a cellular model of Alzheimer's disease.
PMID 25051175
Abstract
Epigenetic modifications, particularly histone acetylation, have been implicated in Alzheimer's disease (AD). While previous studies have suggested that histone hypoacetylation may regulate the expression of genes associated with memory and learning in AD, little is known about histone regulation of AD-related genes such as Presenilin 1(PS1) and beta-site amyloid precursor protein cleaving enzyme 1(BACE1). By utilizing neuroblastoma N2a cells transfected with Swedish mutated human amyloid precursor protein (APP) (N2a/APPswe) and wild-type APP (N2a/APPwt) as cellular models of AD, we examined the alterations of histone acetylation at the promoter regions of PS1 and BACE1 in these cells. Our results revealed that histone H3 acetylation in PS1 and BACE1 promoters is markedly increased in N2a/APPswe cells when compared to N2a/APPwt cells and control cells (vector-transfected), respectively, causing the elevated expression of PS1 and BACE1. In addition, expression of histone acetyltransferase (HAT) adenoviral E1A-associated 300-kDa protein (p300) is dramatically enhanced in N2a/APPswe cells compared to N2a/APPwt and control cells. We have further demonstrated the direct binding of p300 protein to the PS1 and BACE1 promoters in N2a/APPswe cells. The expression levels of H3 acetylation of the PS1 and BACE1 promoters and p300 protein, however, were found to be not significantly different in N2a/APPwt cells when compared to controls in our studies. Furthermore, curcumin, a natural selective inhibitor of p300 in HATs, significantly suppressed the expression of PS1 and BACE1 through inhibition of H3 acetylation in their promoter regions in N2a/APPswe cells. These findings indicated that histone acetyltransferase p300 plays a critical role in controlling the expression of AD-related genes through regulating the acetylation of their promoter regions, suggesting that p300 may represent a novel potential therapeutic target for AD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA003128
Anti-EP300 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004112
Anti-EP300 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502019
Anti-NOTCH1 (Cleaved-Val1754) antibody produced in rabbit, affinity isolated antibody
N4788 Anti-NOTCH1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2101618
Anti-NOTCH1 antibody produced in rabbit, affinity isolated antibody
08511
Curcumin, analytical standard
C21H20O6
00280590
Curcumin, primary pharmaceutical reference standard
C21H20O6
78246
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
C21H20O6
1151855
Curcumin, United States Pharmacopeia (USP) Reference Standard
C21H20O6
WH0002033M2
Monoclonal Anti-EP300 antibody produced in mouse, clone 1D2, purified immunoglobulin, buffered aqueous solution
SAB1400094
Monoclonal Anti-EP300 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
SAB1403776
Monoclonal Anti-EP300 antibody produced in mouse, clone 1D2, ascites fluid
SAB4700742 Monoclonal Anti-Notch 1 antibody produced in mouse, clone mN1A, purified immunoglobulin, buffered aqueous solution
SAB4200024 Monoclonal Anti-Notch 1 antibody produced in rat, ~1.0 mg/mL, clone ICN 5B5, purified immunoglobulin, buffered aqueous solution
N6786
Monoclonal Anti-Notch1 antibody produced in mouse, clone mN1A, purified immunoglobulin, buffered aqueous solution
SAB3300078
Monoclonal Anti-Notch1 antibody produced in mouse, clone 3E12, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402292
Monoclonal Anti-NOTCH1 antibody produced in mouse, clone 4G1, purified immunoglobulin, buffered aqueous solution
PLA0222
Rabbit anti-Notch1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0096
Rabbit anti-p300 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.