EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Determination of ochratoxin A in Capsicum spp. (paprika and chili) by immunoaffinity column cleanup and liquid chromatography: collaborative study.
PMID 25051637
Abstract
A method validation study for the determination of ochratoxin A in Capsicum spp. (paprika and chili) was conducted according to the International Union of Pure and Applied Chemistry harmonized protocol. The method is based on the extraction of samples with aqueous methanol, followed by an immunoaffinity column cleanup. The determination is carried out by RP-HPLC coupled with a fluorescence detector. The study involved 21 participants representing a cross-section of research, private, and official control laboratories from 14 European Union (EU) Member States and Singapore. Mean recoveries reported ranged from 83.7 to 87.5%. The RSD for repeatability (RSDr) ranged from 1.7 to 14.3%. The RSD for reproducibility (RSDR) ranged from 9.1 to 27.5%, reflecting HorRat values from 0.4 to 1.3 according to the Horwitz function modified by Thompson. The correction for recovery of results from naturally contaminated samples further improved the reproducibility of the method. The method showed acceptable within-laboratory and between-laboratory precision for each matrix, and it conforms to requirements set by current EU legislation.