EMAIL THIS PAGE TO A FRIEND
Food chemistry
Instrumental and sensory characterisation of Solaris white wines in Denmark.
PMID 25053038
Abstract
This study aimed to investigate the volatile and non-volatile compositions as well as sensory properties of the most common monovarietal white wine (var. Solaris) in Denmark. Using dynamic headspace sampling (DHS) coupled to gas chromatography-mass spectrometry (GC-MS), 79 volatile compounds were identified. Among the major non-volatile components glycerol, sulphite, sugars and organic acids were analysed. A primary sensory difference was observed among wine samples, half of which were characterised by floral and fruity flavours (peach/apricot, Muscat, melon, banana and strawberry) while the remainder were described by less pleasant flavours, such as chemical, wood and rooibos/smoke. Partial least squares regression (PLS) showed that acetates and ethyl esters of straight-chain fatty acids were associated with floral and fruity odours while ethyl esters of branched-chain fatty acids were less associated with them. The study also suggested that differences in vintage were less characteristic than differences caused due to sulphite management by producers.