EMAIL THIS PAGE TO A FRIEND
Journal of biomedical optics
Microfluidic surface-enhanced Raman scattering sensor with monolithically integrated nanoporous gold disk arrays for rapid and label-free biomolecular detection.
PMID 25054918
Abstract
We present a microfluidic surface-enhanced Raman scattering (SERS) sensor for rapid and label-free biomolecular detection. Our sensor design mitigates a common limiting factor in microfluidic SERS sensors that utilize integrated nanostructures: low-efficiency transport of biomolecules to nanostructured surface which adversely impacts sensitivity. Our strategy is to increase the total usable nanostructured surface area, which provides more adsorption sites for biomolecules. Specifically, a nanoporous gold disk (NPGD) array, a highly effective SERS substrate, has been monolithically integrated inside a microfluidic chip. Individual NPGD is known to feature an order of magnitude larger surface area than its projected disk area. The increased surface area arises from nanoscale pores and ligaments three-dimensionally distributed in the NPGD, which manifest themselves as high-density SERS hot-spots. High-density NPGD arrays further guarantee large coverage of these hot-spots on the microchannel floor. The sensor performance has been demonstrated using Rhodamine 6G to quantify spatial uniformity and determine the shortest detection time. Next, the sensor is applied to detect two biomolecules, dopamine and urea, with unprecedented detection limit and speed compared to other existing microfluidic SERS sensors. The sensor holds great promise in point-of-care applications for various biomolecular detections.
Related Materials