EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Molecularly imprinted polymers for cleanup and selective extraction of curcuminoids in medicinal herbal extracts.
PMID 25056874
Abstract
This paper describes the synthesis of novel molecularly imprinted polymers (MIPs), prepared by a noncovalent imprinting approach, for cleanup and preconcentration of curcumin (CUR) and bisdemethoxycurcumin (BDMC) from medicinal herbal extracts and further analysis by high-performance liquid chromatography with fluorescence detection (HPLC-FLD). Two molecular mimics, a mixture of reduced BDMCs and 4-(4-hydroxyphenyl)-2-butanone (HPB), have been synthesized and applied as templates for MIP synthesis. The polymers were prepared using N-(2-aminoethyl) methacrylamide (EAMA) as functional monomer, ethylene glycol dimethacrylate (EDMA) as the cross-linker (in a 1:5 molar ratio), and a mixture of acetonitrile/dimethylsulfoxide (90%, v/v) as porogen. MIPs prepared using a mixture of reduced BDMCs as template showed higher selectivity for CUR and BDMC than those obtained with HPB, with imprinting factors of 3.5 and 2.7 for CUR and BDMC, respectively, using H2O/acetonitrile (65:35, v/v) as mobile phase. The adsorption isotherms for CUR in the MIP and the nonimprinted polymer (NIP) were fitted to the Freundlich isotherm model, and the calculated average binding affinities for CUR were (17 ± 2) and (8 ± 1) mM(-1) for the MIP and the NIP, respectively. The polymers were packed into solid-phase extraction (SPE) cartridges, and the optimized molecularly imprinted solid-phase extraction (MISPE-HPLC) with fluorescence detection (FLD) method allowed the extraction of both curcuminoids from aqueous samples (50 mM NH4Ac, pH 8.8) followed by a selective washing with acetonitrile/NH4Ac, 50 mM at pH 8.8 (30:70%, v/v), and elution with 3 × 1 mL of MeOH. Good recoveries and precision ranging between 87 and 92%, with relative standard deviation (RSD) of <5.3% (n = 3), were obtained after the preconcentration of 10-mL solutions containing both CUR and BDMC at concentrations in the range of 0-500 μg L(-1). The optimized method has been applied to the analysis of both curcuminoids in medicinal herbal extracts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442840
4,4′-Isopropylidenediphenol, analytical standard
C15H16O2
178519
4-(4-Hydroxyphenyl)-2-butanone, 99%
C10H12O2
68524
4-(4-Hydroxyphenyl)-2-butanone, analytical standard
C10H12O2
W258806
4-(4-Hydroxyphenyl)-2-butanone, ≥98%, FCC, FG
C10H12O2
W258814
4-(4-Hydroxyphenyl)-2-butanone, natural, ≥98%, FCC, FG
C10H12O2
346942
4-Bromo-3,5-dimethylphenyl N-methylcarbamate, 98%
C10H12BrNO2
92264
4-Methoxyphenol, analytical standard
C7H8O2
54050
4-Methoxyphenol, purum, ≥98.0% (HPLC)
C7H8O2
M18655
4-Methoxyphenol, ReagentPlus®, 99%
C7H8O2
N11131
BDMC, analytical standard
C10H12BrNO2
90594
Bisdemethoxycurcumin, analytical standard
C19H16O4
B6938
Bisdemethoxycurcumin, ≥98% (HPLC), solid
C19H16O4
1075305 Bisdesmethoxycurcumin, United States Pharmacopeia (USP) Reference Standard
C19H16O4
42088
Bisphenol A, certified reference material, TraceCERT®
C15H16O2
Y0000012
Hymecromone impurity A, European Pharmacopoeia (EP) Reference Standard
C6H6O2
1598813
Raspberry ketone, United States Pharmacopeia (USP) Reference Standard
C10H12O2
53363
Resorcinol, certified reference material, TraceCERT®
C6H6O2
R5645
Resorcinol, BioXtra, ≥99%
C6H6O2
398047
Resorcinol, ACS reagent, ≥99.0%
C6H6O2
307521
Resorcinol, ReagentPlus®, 99%
C6H6O2
R406
Resorcinol, reagent grade, 98%
C6H6O2
16101
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
C6H6O2
83601
Resorcinol, tested according to Ph.Eur.
C6H6O2
W358908
Resorcinol, ≥98%, FG
C6H6O2
Y0000767
Resorcinol, European Pharmacopoeia (EP) Reference Standard
C6H6O2
1602003
Resorcinol, United States Pharmacopeia (USP) Reference Standard
C6H6O2
86854
Tetrabutylammonium hydroxide solution, ~40% in water, for ion chromatography
C16H37NO
86863
Tetrabutylammonium hydroxide solution, 54.0-56.0% in H2O
C16H37NO
86880
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
C16H37NO
178780
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
C16H37NO