EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Frontal cortical and subcortical projections provide a basis for segmenting the cingulum bundle: implications for neuroimaging and psychiatric disorders.
PMID 25057206
Abstract
The cingulum bundle (CB) is one of the brain's major white matter pathways, linking regions associated with executive function, decision-making, and emotion. Neuroimaging has revealed that abnormalities in particular locations within the CB are associated with specific psychiatric disorders, including depression and bipolar disorder. However, the fibers using each portion of the CB remain unknown. In this study, we used anatomical tract-tracing in nonhuman primates (Macaca nemestrina, Macaca fascicularis, Macaca mulatta) to examine the organization of specific cingulate, noncingulate frontal, and subcortical pathways through the CB. The goals were as follows: (1) to determine connections that use the CB, (2) to establish through which parts of the CB these fibers travel, and (3) to relate the CB fiber pathways to the portions of the CB identified in humans as neurosurgical targets for amelioration of psychiatric disorders. Results indicate that cingulate, noncingulate frontal, and subcortical fibers all travel through the CB to reach both cingulate and noncingulate targets. However, many brain regions send projections through only part, not all, of the CB. For example, amygdala fibers are not present in the caudal portion of the dorsal CB. These results allow segmentation of the CB into four unique zones. We identify the specific connections that are abnormal in psychiatric disorders and affected by neurosurgical interventions, such as deep brain stimulation and cingulotomy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB152
Anti-Tyrosine Hydroxylase Antibody, 100 µL, Anti-Tyrosine Hydroxylase Antibody detects level of TH and has been published and validated for use in ELISA, IF, IH, IH(P), IP and WB.
SAB2104059
Anti-BLVRB antibody produced in rabbit, affinity isolated antibody
HPA041698
Anti-BLVRB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA041937
Anti-BLVRB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406997
Anti-FST antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018155
Anti-FST antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV42287
Anti-FST antibody produced in rabbit, IgG fraction of antiserum
SAB2100858
Anti-FST antibody produced in rabbit, affinity isolated antibody
HPA051298
Anti-GBGT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406446
Anti-SLC4A1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA015584
Anti-SLC4A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33801
Anti-SLC4A1 antibody produced in rabbit, affinity isolated antibody
WH0000645M9
Monoclonal Anti-BLVRB antibody produced in mouse, clone 2F4, purified immunoglobulin, buffered aqueous solution
SAB1404832
Monoclonal Anti-FST antibody produced in mouse, clone 4B11, purified immunoglobulin, buffered aqueous solution
SAB1402515
Monoclonal Anti-FST antibody produced in mouse, clone 1G4, ascites fluid, solution
SAB1402919
Monoclonal Anti-SLC4A1 antibody produced in mouse, clone 2D5, purified immunoglobulin, buffered aqueous solution