EMAIL THIS PAGE TO A FRIEND
Journal of immunological methods
Comparative characterization of mAb producing hapten-specific hybridoma cells by flow cytometric analysis and ELISA.
PMID 25058593
Abstract
A novel method that optimizes the screening for antibody-secreting hapten-specific hybridoma cells by using flow cytometry is described. Cell clones specific for five different haptens were analyzed. We selectively double stained and analyzed fixed hybridoma cells with fluorophore-labeled haptens to demonstrate the target-selectivity, and with a fluorophore-labeled anti-mouse IgG antibody to characterize the level of surface expression of membrane-bound IgGs. ELISA measurements with the supernatants of the individual hybridoma clones revealed that antibodies from those cells, which showed the highest fluorescence intensities in the flow cytometric analysis, also displayed the highest affinities for the target antigens. The fluorescence intensity of antibody-producing cells corresponded well with the produced antibodies' affinities toward their respective antigens. Immunohistochemical staining verified the successful double labeling of the cells. Our method makes it possible to perform a high-throughput screening for hybridoma cells, which have both an adequate IgG production rate and a high target affinity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15241
N,O-Bis(trimethylsilyl)acetamide, purum, ≥95.0% (GC)
C8H21NOSi2