EMAIL THIS PAGE TO A FRIEND
Molecular endocrinology (Baltimore, Md.)
ErbB4, a receptor tyrosine kinase, coordinates organization of the seminiferous tubules in the developing testis.
PMID 25058600
Abstract
Although close to every fifth couple nowadays has difficulty conceiving, the molecular mechanisms behind the decline in human reproduction remain poorly understood. We report here that the receptor tyrosine kinase Erbb4 is a candidate causal gene, because it is expressed in a sexually dimorphic manner and is abundant in the developing and adult testes in the mouse. Sertoli cell-specific Erbb4-knockout mice have a compromised 3-dimensional organization of the testicular seminiferous tubules that affects their fertility. More specifically, adhesion defects are observed in the absence of Erbb4, which are characterized by changes in the expression of laminin-1, N-cadherin, claudin-3, and certain cell-cell junction components between the Sertoli and germ cells. Interestingly, Erbb4 knockout also had an effect on the Leydig cells, which suggests a paracrine influence of Sertoli cells expressing ErbB4. Many of the defects observed in Erbb4-knockout mice are rescued in targeted ERBB4 gain-of-function mice, pointing to a coordination role for ErbB4 in the developing testis. Thus, the ErbB4 receptor tyrosine kinase promotes seminiferous tubule development by controlling Sertoli cell and germ cell adhesion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1305442
ANTI-AMH (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100114
Anti-AMH antibody produced in rabbit, affinity isolated antibody
SAB1405763
Anti-EPHB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4500732
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB4500775
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB4500776
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB1412026
ANTI-EPHB2 antibody produced in mouse, clone 1F10, ascites fluid, buffered aqueous solution
SAB2502079
Anti-ESR2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV31652
Anti-ESR2 antibody produced in rabbit, affinity isolated antibody
SAB2100713
Anti-ESR2 antibody produced in rabbit, affinity isolated antibody
SAB2105299
Anti-ESR2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4300324
Anti-GATA1 (Ab-142) antibody produced in rabbit, affinity isolated antibody
SAB4300325
Anti-GATA1 (Ab-310) antibody produced in rabbit, affinity isolated antibody
SAB2500452
Anti-GATA1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA000232
Anti-GATA1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100897
Anti-GATA1 antibody produced in rabbit, affinity isolated antibody
G0290 Anti-GATA1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA000233
Anti-GATA1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
G4671 Anti-GATA1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4501122
Anti-GATA1 antibody produced in rabbit, affinity isolated antibody
AV50183
Anti-Siglec-12 (AB1) antibody produced in rabbit, affinity isolated antibody
AV50235
Anti-Siglec-12 (AB2) antibody produced in rabbit, affinity isolated antibody
AV50236
Anti-Siglec-12 (AB3) antibody produced in rabbit, affinity isolated antibody
SAB1402765
Monoclonal Anti-EPHB1 antibody produced in mouse, clone 4G6, purified immunoglobulin, buffered aqueous solution
SAB5300309 Monoclonal Anti-EPHB1 antibody produced in mouse, clone 5F10A4, ascites fluid
WH0002048M3
Monoclonal Anti-EPHB2 antibody produced in mouse, clone 4D1, purified immunoglobulin, buffered aqueous solution
SAB1403781
Monoclonal Anti-EPHB2 antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
SAB5300480
Monoclonal Anti-EPHB2 antibody produced in mouse, clone 2D12C6, ascites fluid
WH0002100M1
Monoclonal Anti-ESR2 antibody produced in mouse, clone 3F3, purified immunoglobulin, buffered aqueous solution
SAB2702145
Monoclonal Anti-ESR2 antibody produced in mouse