EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Scalable high-fidelity growth of semiconductor nanorod arrays with controlled geometry for photovoltaic devices using block copolymers.
PMID 25059670
Abstract
Controlled density semiconducting oxide arrays are highly desirable for matching nanometer length scales specific to emerging applications. This work demonstrates a facile one-step method for templating hydrothermal growth which provides arrays with high-fidelity tuning of nanorod spacing and diameter. This solution-based method leverages the selective swelling of block copolymer micelle templates, which can be rationally designed by tuning molecular weight and volume fraction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284467
2-Methoxyethanol, anhydrous, 99.8%
C3H8O2
270482
2-Methoxyethanol, for HPLC, ≥99.9%
C3H8O2
360503
2-Methoxyethanol, contains 50 ppm BHT as stabilizer, ACS reagent, ≥99.3%
C3H8O2
185469
2-Methoxyethanol, ReagentPlus®, ≥99.0%, contains 50 ppm BHT as stabilizer
C3H8O2
64719
2-Methoxyethanol, for amino acid analysis, ≥99.5% (GC)
C3H8O2
88907
2-Methoxyethanol, analytical standard
C3H8O2
33457
2-Methoxyethanol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
C3H8O2
PHR1544
2-Methoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O2
55561
Aucubin, analytical standard
C15H22O9
00090590
Aucubin, primary pharmaceutical reference standard
C15H22O9
02400
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
C2H7NO
00278
Ethanolamine, analytical standard
C2H7NO
02412
Ethanolamine solution, for peptide synthesis, ~74% (T), ~70% in methanol
C2H7NO
H6404
Hexamethylenetetramine, BioXtra
C6H12N4
33233
Hexamethylenetetramine, puriss. p.a., Reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
C6H12N4
15614
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
C6H12N4
55321
Hexamethylenetetramine, analytical reference material
C6H12N4
398160
Hexamethylenetetramine, ACS reagent, ≥99.0%
C6H12N4
H11300
Hexamethylenetetramine, ReagentPlus®, 99%
C6H12N4
Y0001184
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard
C2H7NO