EMAIL THIS PAGE TO A FRIEND
Journal of viral hepatitis
Role of CYP27B1+2838 promoter polymorphism in the treatment of chronic hepatitis B HBeAg negative with PEG-interferon.
PMID 25060765
Abstract
In HBV-infected patients, the vitamin D deficiency has been related to chronic liver diseases, progression of hepatic fibrosis and poor response to the treatment. The CYP27B1 gene, which encodes the 1-α-hidroxylase and involved in the 1,25-dihydroxyvitamin D synthesis, was recently associated to type-1 diabetes, autoimmune disorders and treatment response in HCV. Then, we aimed to investigate the role of CYP27B1 polymorphisms in HBV treatment with PEG-IFN. We retrospectively enrolled 190 patients with chronic hepatitis B HBeAg negative treated for 48 weeks with PEG-IFN α-2a. We examined the role of rs4646536 CYP27B1 SNP (CYP27B1+2838) according to virological and serological response. Our results showed that the TT genotype of CYP27B1+2838 was significantly prevalent in patients with end-of-therapy virological response (37.6%) vs CT/CC (9.4%) (P < 0.001). Virological relapse was prevalent in patients with CT/CC genotype (12.6%) vs TT genotype (2.1%) (P < 0.001). TT genotype was also related to HBsAg loss (P = 0.004) and anti-HBs appearance (P = 0.002). In the multivariate analysis, the TT genotype resulted to be a good positive predictor of sustained virological response (OR = 5.632, IC = 1.938-16.368, P = 0.001) and serological response (OR = 6.161, IC = 1.856-20.457, P = 0.003). The CYP27B1+2838 polymorphism may be useful as pretreatment factor to selection of patients with higher probability of response to therapy.