EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Effects of recombinant activins on steroidogenesis in human granulosa-lutein cells.
PMID 25062451
Abstract
Exerting a broad range of biological effects in various tissues, activins are homo- or heterodimers of activin/inhibin β-subunits (βA, βB, βC, and βE in humans). Although activins A (βAβA), B (βBβB), AB (βAβB), and AC (βAβC) have been demonstrated in the female reproductive system, little is known about their individual functions in the ovary. To investigate the biological roles and activities of activins in regulating steroidogenesis in human granulosa cells. Human granulosa-lutein cells obtained from 32 patients undergoing in vitro fertilization were used to investigate the effects of activin A, B, AB, and AC on the expression of steroidogenic enzymes and steroid production. An academic research center. mRNA and protein levels were examined by reverse transcription quantitative real-time PCR and Western blot analysis, respectively. The production of estradiol and progesterone was measured by enzyme immunoassay. P450 aromatase, FSH receptor, and estradiol levels were increased, whereas steroidogenic acute regulatory protein (StAR), LH receptor, and progesterone levels were decreased after treatment with activin A, B, and AB, but not activin AC. FSH or LH induced the production of aromatase/estradiol and StAR/progesterone; however, pretreatment with activin A, B, or AB enhanced the effects of gonadotropins on aromatase/estradiol, but suppressed their effects on StAR/progesterone. Treatment with activin A, B, or AB induced the phosphorylation of SMA- and MAD-related proteins (SMAD2 and 3), whereas activin AC had no such effects. Furthermore, co-culture of activin AC (1-100 ng/mL) with activin A (25 ng/mL) did not alter the effects of activin A on P450 aromatase or StAR mRNA levels. Activin A, B, and AB have similar effects on steroidogenesis in human granulosa cells. In contrast, activin AC is not biologically active and does not act as a competitive antagonist.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4362
Activin A active human, Animal-component free, recombinant, expressed in Nicotiana, >97% (SDS-PAGE)
A4941 Activin A human, >95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, lyophilized powder, cell culture tested
H4666
Activin A human, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
SRP6153
Activin A human, recombinant, expressed in human cells, ≥95% (SDS-PAGE)
SRP3003 Activin A human, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC), cell culture tested
HPA051194
Anti-CYP19A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410280
Anti-CYP19A1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500606
Anti-Cytochrome P450 19A1 antibody produced in rabbit, affinity isolated antibody
SAB4504210
Anti-phospho-Smad3 (pSer204) antibody produced in rabbit, affinity isolated antibody
SAB4504211
Anti-phospho-Smad3 (pSer208) antibody produced in rabbit, affinity isolated antibody
SAB4504212
Anti-phospho-Smad3 (pSer213) antibody produced in rabbit, affinity isolated antibody
SAB4503781
Anti-phospho-Smad3 (pSer425) antibody produced in rabbit, affinity isolated antibody
SAB4300253
Anti-phospho-SMAD3 (pSer425) antibody produced in rabbit, affinity isolated antibody
SAB4504209
Anti-phospho-Smad3 (pThr179) antibody produced in rabbit, affinity isolated antibody
SAB4300564
Anti-SMAD3 (Ab-208) antibody produced in rabbit, affinity isolated antibody
SAB4300565
Anti-SMAD3 (Ab-425) antibody produced in rabbit, affinity isolated antibody
AV100621
Anti-SMAD3 antibody produced in rabbit, IgG fraction of antiserum
SAB4200342
Anti-SMAD3 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2702052
Anti-SMAD3 antibody produced in rabbit
SAB4501811
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
SAB4501812
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
SAB4501813
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
WH0001588M3
Monoclonal Anti-CYP19A1 antibody produced in mouse, clone 2B6, purified immunoglobulin, buffered aqueous solution
WH0004088M1
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 2C12, purified immunoglobulin, buffered aqueous solution
WH0004088M5 Monoclonal Anti-SMAD3 antibody produced in mouse, clone 4D5, purified immunoglobulin, buffered aqueous solution
SAB1400160
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 7F3, purified immunoglobulin, buffered aqueous solution
SAB1404037
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 4F9, purified immunoglobulin, buffered aqueous solution
1568007
Progesterone, United States Pharmacopeia (USP) Reference Standard
C21H30O2
46665
Progesterone, VETRANAL, analytical standard
C21H30O2
PHR1142
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
C21H30O2