EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Linolenic acid provides multi-cellular protective effects after photothrombotic cerebral ischemia in rats.
PMID 25062759
Abstract
Alpha-linolenic acid (LIN) has been shown to provide neuroprotective effects against cerebral ischemia. LIN is a potent activator of TREK-1 channel and LIN-induced neuroprotection disappears in Trek1-/- mice, suggesting that this channel is directly related to the LIN-induced resistance of brain against ischemia. However, the cellular mechanism underlying LIN induced neuroprotective effects after ischemia remains unclear. In this study, using a rat photochemical brain ischemia model, we investigated the effects of LIN on the protein abundance of astrocytic glutamate transporter and AQP4, microglia activation, cell apoptosis and behavioral recovery following ischemia. Administration of LIN rescued the protein abundance of astrocytic glutamate transporter GLT-1, decreased the protein abundance of AQP4 and brain edema, inhibited microglia activation, attenuated cell apoptosis and improved behavioral function recovery. Meanwhile, TREK-1 was widely distributed in the cortex and hippocampus, primarily localized in astrocytes and neurons. LIN could potentiate the TREK-1 mediated astrocytic passive conductance and hyperpolarize the membrane potential. Our results suggest that LIN provides multiple cellular neuroprotective effects in cerebral ischemia. TREK-1 may serve as a promising multi-mechanism therapeutic target for the treatment of stroke.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA049234
Anti-AIF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA009172
Anti-EAAT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA056054
Anti-KCNK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
F1262 Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F6005 Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
F7512 Anti-Rabbit IgG (whole molecule)−FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
SAB2102171
Anti-SLC1A2 antibody produced in rabbit, affinity isolated antibody
SAB2104141
Anti-SLC1A2 antibody produced in rabbit, affinity isolated antibody
SAB1411282
Anti-SLC1A2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
T6448
Anti-TREK-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
L2376
Linolenic acid, ≥99%
C18H30O2
62170
Linolenic acid, ~70% (GC)
C18H30O2
62160
Linolenic acid, analytical standard
C18H30O2
AMAB90537
Monoclonal Anti-AQP4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0178, purified immunoglobulin, buffered aqueous glycerol solution