EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Genomic characterization and transcriptional studies of the starch-utilizing strain Bifidobacterium adolescentis 22L.
PMID 25063659
Abstract
Bifidobacteria are members of the gut microbiota, but the genetic basis for their adaptation to the human gut is poorly understood. The analysis of the 2,203,222-bp genome of Bifidobacterium adolescentis 22L revealed a nutrient acquisition strategy that targets diet/plant-derived glycans, in particular starch and starch-like carbohydrates. Starch-like carbohydrates were shown to support the growth of B. adolescentis 22L. Transcriptome profiling of 22L cultures grown under in vitro conditions or during colonization of the murine gut by RNA sequencing and quantitative real-time PCR assays revealed the expression of a set of chromosomal loci responsible for starch metabolism as well as for pilus production. Such extracellular structures include so-called sortase-dependent and type IVb pili, which may be involved in gut colonization of 22L through adhesion to extracellular matrix proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0750
D-(+)-Galactose, ≥99%
C6H12O6
G0625
D-(+)-Galactose, ≥98%
C6H12O6
G6404
D-(+)-Galactose, BioXtra, pH 5.0-7.0 (20 °C, 1 M in H2O), ≥99%
C6H12O6
G5388
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
C6H12O6
48260
D-(+)-Galactose, for microbiology, ≥99.0%
C6H12O6
15522
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
C6H12O6
ARK2181
D-(+)-Mannose, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C6H12O6
M2069
D-(+)-Mannose, from wood, ≥99%
C6H12O6
M8574
D-(+)-Mannose, synthetic, ≥99%
C6H12O6
M6020
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
C6H12O6
63579
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
63580
D-(+)-Mannose, for microbiology, ≥99%
C6H12O6
63582
D-(+)-Mannose, for microbiology, ≥99.0% (sum of enantiomers, HPLC)
C6H12O6
M4625
D-(+)-Mannose
C6H12O6
63560
D-Mannitol, ACS reagent, for microbiology, ≥99.0%
C6H14O6
17311
D-Mannitol, tested according to Ph.Eur.
C6H14O6
78513
D-Mannitol, ≥99.9999% (metals basis), for boron determination
C6H14O6
W360600
D-(+)-Xylose, ≥99%
C5H10O5
X1500
D-(+)-Xylose, ≥99%
C5H10O5
X3877
D-(+)-Xylose, BioXtra, ≥99%
C5H10O5
95729
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
C5H10O5
47253
D-(+)-Xylose, analytical standard
C5H10O5
1287700 Galactose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
PHR1206
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
G0050000
Galactose, European Pharmacopoeia (EP) Reference Standard
C6H12O6
ARK2176
Galactose, plant-derived, PharmaGrade, USP/NF, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C6H12O6
17814
Lactose, tested according to Ph.Eur.
C12H22O11
L7877
Lactulose, ≥95%
C12H22O11
1356803
Lactulose, United States Pharmacopeia (USP) Reference Standard
C12H22O11
L0130000
Lactulose, European Pharmacopoeia (EP) Reference Standard
C12H22O11