EMAIL THIS PAGE TO A FRIEND
Atherosclerosis
Plasma total homocysteine and carotid intima-media thickness in type 1 diabetes: a prospective study.
PMID 25063949
Abstract
Plasma total homocysteine (tHcy) has been positively associated with carotid intima-media thickness (IMT) in non-diabetic populations and in a few cross-sectional studies of diabetic patients. We investigated cross-sectional and prospective associations of a single measure of tHcy with common and internal carotid IMT over a 6-year period in type 1 diabetes. tHcy levels were measured once, in plasma obtained in 1997-1999 from patients (nxa0=xa0599) in the Epidemiology of Diabetes Interventions and Complications (EDIC) study, the observational follow-up of the Diabetes Control and Complications Trial (DCCT). Common and internal carotid IMT were determined twice, in EDIC "Year 6" (1998-2000) and "Year 12" (2004-2006), using B-mode ultra-sonography. After adjustment, plasma tHcy [median (interquartile range): 6.2 (5.1, 7.5) μmol/L] was significantly correlated with age, diastolic blood pressure, renal dysfunction, and smoking (all pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69453
L-Homocysteine, ≥98.0% (NT)
C4H9NO2S