EMAIL THIS PAGE TO A FRIEND
Circulation. Cardiovascular genetics
Functional analysis of single-nucleotide polymorphisms in the regulation of coactivator-associated arginine methyltransferase 1 expression and plasma homocysteine levels.
PMID 25064859
Abstract
Hyperhomocysteinemia is a risk factor for cardiovascular disease. Coactivator-associated arginine methyltransferase 1 (CARM1) participates in the synthesis of homocysteine, but whether the genetic variations regulate CARM1 expression and homocysteine levels remains unknown. Functional analyses combined with an association study were conducted to identify the causal variant for CARM1 expression and homocysteine levels. Based on functional annotations obtained from Encyclopedia of DNA Elements, we selected 4 potentially functional single-nucleotide polymorphisms in the CARM1 gene and investigated their effect on CARM1 transcription levels in vivo. rs117569851, located in the promoter region of CARM1, as well as rs12460421 and rs4804544, was associated with CARM1 expression levels, and the last 2 single-nucleotide polymorphisms were discovered in high linkage disequilibrium with rs117569851 (r(2)=0.9 and 1.0) in our study sample. rs117569851 was further identified to be responsible for regulating CARM1 expression. The T allele disrupted the binding of early growth response-1, which led to the downregulation of transcriptional activity in vitro and CARM1 mRNA levels in vivo. In addition, rs117569851 was associated with plasma homocysteine levels in a Chinese population (n=406), with a 2.16 μmol/L decrease per copy of T allele. The present study suggests that a noncoding variant in the CARM1-promoter functions as a regulator of gene transcription and homocysteine levels.