EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
A novel UPLC-MS/MS method for sensitive quantitation of boldine in plasma, a potential anti-inflammatory agent: application to a pharmacokinetic study in rats.
PMID 25065486
Abstract
Boldine is a potential anti-inflammatory agent found in several different plants. Published bioanalytical methods using HPLC with ultraviolet and fluorescent detection lacked enough sensitivity and required tedious sample preparation procedures. Herein, we describe the development of a novel ultra-high performance LC with MS/MS for determination of boldine in plasma. Boldine in plasma was recovered by liquid-liquid extraction using 1 mL of methyl tert-butyl ether. Chromatographic separation was performed on a C18 column at 45°C, with a gradient elution consisting of acetonitrile and water containing 0.1% (v/v) formic acid at a flow rate of 0.3 mL/min. The detection was performed on an electrospray triple-quadrupole MS/MS by positive ion multiple reaction monitoring mode. Good linearity (r(2) > 0.9926) was achieved in a concentration range of 2.555-2555 ng/mL with a lower limit of quantification of 2.555 ng/mL for boldine. The intra- and inter-day precisions of the assay were 1.2-6.0 and 1.8-7.4% relative standard deviation with an accuracy of -6.0-8.0% relative error. This newly developed method was successfully applied to a single low-dose pharmacokinetic study in rats and was demonstrated to be simpler and more sensitive than the published methods, allowing boldine quantification in reduced plasma volume.