EMAIL THIS PAGE TO A FRIEND
Canadian journal of physiology and pharmacology
Acute arsenic treatment alters arachidonic acid and its associated metabolite levels in the brain of C57Bl/6 mice.
PMID 25065748
Abstract
The toxic effects of arsenic on the whole brain, as well as the discrete regions, has been previously reported for mice. We investigated the effects of acute arsenite (As(III)) on brain levels of arachidonic acid (AA) and its associated metabolites generated through cytochrome P450 (CYP), cyclooxygenase (COX), and lipoxygenase (LOX) pathways. Our results demonstrated that acute As(III) treatment (12.5 mg·(kg body mass)(-1)) decreases cytosolic phospholipase A2 (cPLA2) with a subsequent decrease in its catalytic activity and brain AA levels. In addition, As(III) differentially altered CYP epoxygenases and CYP ω-hydroxylases, but it did not affect brain Ephx2 mRNA or sEH catalytic activity levels. As(III)-mediated effects on Cyps caused an increase in brain 5,6-epoxyeicosatrienoic acid (5,6-EET) and 16/17-hydroxyeicosatetreinoic acid (16/17-HETE) levels, and a decrease in 18- and 20-HETE levels. Furthermore, As(III) increased cyclooxygenase-2 (COX-2) mRNA while decreasing prostaglandins F2α (PGF2α) and PGJ2. As(III) also increased brain 5-lipoxygenase (5-LOX) and 15-LOX mRNA, but decreased 12-LOX mRNA. These changes in LOX mRNA were associated with a decrease in 8/12-HETE levels only. In conclusion, this is the first demonstration that As(III) decreases AA levels coinciding with alterations to EET, HETE, and PG levels, which affects brain development and neurochemistry.