EMAIL THIS PAGE TO A FRIEND
Chirality
Enzymatic hydrolysis of esters containing a tetrazole ring.
PMID 25065812
Abstract
The lipase-catalyzed enantioselective hydrolysis of acetates containing tetrazole moiety was studied. Among all tested lipases, Novozyme SP 435 allowed to obtain optically active 4-(5-aryl-2H-tetrazol-2yl)butan-2-ol and 1-(5-aryl-2H-tetrazol-2yl)-propan-2-ol and their acetates with the highest optical purities (ee = 95%-99%) and excellent enantioselectivity (E>100). Some of the synthesized tetrazole derivatives were screened for their antifungal activity. Racemic mixtures of 4-[5-(4-chlorophenyl)-2H-tetrazol-2-yl)butan-2-ol as well as pure enantiomers of this compound showed promising antifungal activity against F. sambucinum, F. oxysporum, C. coccodes, and A. niger.