EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
New celecoxib multiparticulate systems to improve glioblastoma treatment.
PMID 25066075
Abstract
Treatment of malignant gliomas consists of resection followed by radiotherapy and chemotherapy. Celecoxib (CXB), a selective COX-2 inhibitor, is able to control inflammation and pain, to improve the efficacy of radiotherapy, and to inhibit at high doses the growth of cancer cells. Two new delivery systems for CXB are developed: microspheres (MPs) for implantation in the brain after partial/complete removal of the tumor, and nanoparticles (NPs) for their potential to cross the blood brain barrier and deliver CXB into the CNS. Cell culture assays performed in PC12, SKN-AS and U373-MG cells demonstrate the antiproliferative affects of CXB, with EC50 values of 99.81 μM and 82.4 μM in U373-MG and SKN-AS cells. Encapsulation efficacy of CXB in formulation MP2 (20% CXB) was 74.6 ± 2.2% with a zero-order release rate of 47.8 μg/day/20mg microspheres for 34 days. Uncoated and polysorbate 80-coated CXB-NPs are prepared by nanoprecipitation. Mean sizes of uncoated and coated CXB-NPs were 173.6 ± 44.9 nm and 100.6 ± 62.1 nm. Cerebral cortex images showed a marked increase of fluorescence when the surfactant-coated NPs were administered to rats. These results suggest that both CXB formulations (MPs and NPs) are adequate systems to enhance the effects of chemotherapy in the treatment of malignant brain tumor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001445
Celecoxib, European Pharmacopoeia (EP) Reference Standard
C17H14F3N3O2S