EMAIL THIS PAGE TO A FRIEND
Iranian Red Crescent medical journal
Long-term neurodevelopmental outcomes after preterm birth.
PMID 25068052
Abstract
All over the the world, preterm birth is a major cause of death and important neurodevelopmental disorders. Approximately 9.6% (12.9 million) births worldwide are preterm. In this review, databases such as PubMed, EMBASE, ISI, Scopus, Google Scholar and Iranian databases including Iranmedex, and SID were researched to review relevant literature. A comprehensive search was performed using combinations of various keywords. Cerebral palsy especially spastic diplegia, intellectual disability, visual (retinopathy of prematurity) and hearing impairments are the main neurodevelopmental disorders associated with prematurity. The increased survival of preterm infants was not associated with lower complications. There is now increasing evidence of sustained adverse outcomes into school age and adolescence, for preterm infants.