EMAIL THIS PAGE TO A FRIEND
Journal of endodontics
EZH2, a potential regulator of dental pulp inflammation and regeneration.
PMID 25069920
Abstract
Dental pulp has limited capability to regenerate, which happens in the early stage of pulpitis. An ambiguous relationship exists; inflammation may impair or support pulp regeneration. Epigenetics, which is involved in cell proliferation and inflammation, could regulate human dental pulp cell (HDPCs) regeneration. The aim of this study was to determine the role of the epigenetic mark, enhancer of zeste homolog 2 (EZH2), in the inflammation, proliferation, and regeneration of dental pulp. We used trimethylated histone H3 lysine 27(H3K27me3) and its lysine demethylase 6B (KDM6B) to monitor functional effects of altered EZH2 levels. We detected epigenetic marks (EZH2, H3K27me3, and KDM6B) in pulp tissue by immunohistochemistry and immunofluorescence. EZH2 levels in HDPCs in inflammatory responses or differentiation were analyzed by quantitative polymerase chain reaction and Western blot. Quantitative polymerase chain reaction was used to assess the effects of EZH2 inhibition on interleukins in HDPCs upon tumor necrosis factor alpha stimulation. Cell proliferation was tested by cell counting kit-8, cell cycle, and apoptosis analysis. HDPC differentiation was investigated by quantitative polymerase chain reaction, alkaline phosphatase activity, and oil red O staining. EZH2 and H3K27me3 were decreased, whereas KDM6B was increased in infected pulp tissue and cells, which were similar to HDPC differentiation. EZH2 inhibition suppressed IL-1b, IL-6, and IL-8 messenger RNA (mRNA) in HDPCs upon inflammatory stimuli and impeded HDPC proliferation by decreasing cell number, arresting cell cycle, and increasing apoptosis. Suppressed EZH2 impaired adipogenesis, peroxisome proliferator-activated receptor r (PPAR-r), and CCAAT-enhancer binding protein a (CEBP/a) mRNA in adipogenic induction while enhancing alkaline phosphatase activity, Osx, and bone sialoprotein (BSP) mRNA in mineralization induction of HDPCs. EZH2 inhibited HDPC osteogenic differentiation while enhancing inflammatory response and proliferation, suggesting its role in pulp inflammation, proliferation, and regeneration.