EMAIL THIS PAGE TO A FRIEND
Transplant international : official journal of the European Society for Organ Transplantation
Lung transplantation from donors outside standard acceptability criteria--are they really marginal?
PMID 25070600
Abstract
Lung transplantation (LTx) from "extended donor criteria" donors may reduce significantly organ shortage. However, its influence on results remains unclear. In this study, we evaluate retrospectively the results of LTx from donors outside standard criteria: PaO2/FiO2 ratioxa0xa020 pack-years. Two hundred and forty-eight patients underwent first time LTx in our institution between January 2007 and January 2013. Seventy-nine patients (Group I) received organs from "extended donor criteria" and 169 patients (Group II) from "standard donor criteria." Recipients' and donors' demographics, perioperative variables, and outcome were compared. Donors from Group I were significantly older [median (interquartile range)]: 52.5 (44;58) vs. 42 (28.5;48.5) years (Pxa0