EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Ovarian xenobiotic biotransformation enzymes are altered during phosphoramide mustard-induced ovotoxicity.
PMID 25070981
Abstract
The anti-neoplastic prodrug, cyclophosphamide, requires biotransformation to phosphoramide mustard (PM), which partitions to volatile chloroethylaziridine (CEZ). PM and CEZ are ovotoxicants, however their ovarian biotransformation remains ill-defined. This study investigated PM and CEZ metabolism mechanisms through the utilization of cultured postnatal day 4 (PND4) Fisher 344 (F344) rat ovaries exposed to vehicle control (1% dimethyl sulfoxide (DMSO)) or PM (60μM) for 2 or 4 days. Quantification of mRNA levels via an RT(2) profiler PCR array and target-specific RT-PCR along with Western blotting found increased mRNA and protein levels of xenobiotic metabolism genes including microsomal epoxide hydrolase (Ephx1) and glutathione S-transferase isoform pi (Gstp). PND4 ovaries were treated with 1% DMSO, PM (60μM), cyclohexene oxide to inhibit EPHX1 (CHO; 2mM), or PM + CHO for 4 days. Lack of functional EPHX1 increased PM-induced ovotoxicity, suggesting a detoxification role for EPHX1. PND4 ovaries were also treated with 1% DMSO, PM (60μM), BSO (Glutathione (GSH) depletion; 100μM), GEE (GSH supplementation; 2.5mM), PM ± BSO, or PM ± GEE for 4 days. GSH supplementation prevented PM-induced follicle loss, whereas no impact of GSH depletion was observed. Lastly, the effect of ovarian GSH on CEZ liberation and ovotoxicity was evaluated. Both untreated and GEE-treated PND4 ovaries were plated adjacent to ovaries receiving PM + GEE or PM + BSO treatments. Less CEZ-induced ovotoxicity was observed with both GEE and BSO treatments indicating reduced CEZ liberation from PM. Collectively, this study supports ovarian biotransformation of PM, thereby influencing the ovotoxicity that ensues.