EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological and toxicological methods
Pharmacokinetic-pharmacodynamic (PK-PD) modeling and simulation of 5-fluorouracil for erythropenia in rats.
PMID 25072509
Abstract
The aim of the present study was to develop a simple pharmacokinetic-pharmacodynamic (PK-PD) model in rats that could predict the onset and degree of erythropenia, a severely toxic side effect that severely limits the use of the anticancer agent 5-fluorouracil (5-FU). Total erythrocyte counts, hemoglobin (Hb) concentrations, and hematocrit (Hct) levels were measured in rats following the intravenous bolus administration of 5-FU for 4 days in order to obtain data for an analysis of the PK-PD model. Our PK-PD model consisted of a two-compartment PK model, with two compartments for the PD model and nine structural PK-PD model parameters. After the intravenous bolus administration of 5, 10, or 20 mg/kg of 5-FU to rats, absolute erythrocyte counts, Hb concentrations, and Hct levels transiently decreased, reached minimum levels on Days 7-14, and then returned to baseline levels. The nadir values (Cnadir) for rats treated with 5, 10, or 20 mg/kg of 5-FU were significantly decreased to approximately 79.4, 76.3, or 46.5% of the baseline value (Cbaseline) in erythrocyte counts, 86.3, 83.3, or 45.7% of Cbaseline in Hb concentrations, 88.6, 85.5, or 47.1% of Cbaseline in Hct levels, respectively. The PK-PD model effectively captured the features of erythropenia and Cnadir after 5-FU chemotherapy. This PK-PD model was successfully used to characterize the learner relationship between the area under the plasma 5-FU concentration-time curve (AUC0-∞) following the intravenous bolus administration of 5-FU and the Cnadir in erythrocyte counts, Hb concentrations, and Hct levels after the 5-FU treatment. The results of the present study suggest that the administration of a pharmacokinetically modified dose of 5-FU could minimize the Cnadir in erythrocyte counts, Hb concentrations, and Hct levels following the administration of 5-FU. The PK-PD model and simulation represent valuable approaches for quantifying and predicting erythropenia as well as determining individual doses and the time at which the subsequent course of the treatment should start.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852473
5-Bromouracil, 98%
C4H3BrN2O2