EMAIL THIS PAGE TO A FRIEND
Journal of animal physiology and animal nutrition
Gender-specific effects of a phytogenic feed additive on performance, intestinal physiology and morphology in broiler chickens.
PMID 25073418
Abstract
To date, most studies published were carried out on broilers of the same sex, and possible gender-specific effects of phytogenic substances have not been investigated so far. A 3xa0×xa02 factorial study was performed to examine gender-specific effects of a PFA at two dietary levels (150, 1500xa0ppm) on growth performance, carcass traits and gastrointestinal attributes in broiler chickens versus an untreated control group. The addition of 150xa0ppm of the PFA led to a downregulation of trypsinogen mRNA in pancreas compared with the control group (pxa0