EMAIL THIS PAGE TO A FRIEND
Scientific reports
A tool kit for rapid cloning and expression of recombinant antibodies.
PMID 25073855
Abstract
Over the last four decades, molecular cloning has evolved tremendously. Efficient products allowing assembly of multiple DNA fragments have become available. However, cost-effective tools for engineering antibodies of different specificities, isotypes and species are still needed for many research and clinical applications in academia. Here, we report a method for one-step assembly of antibody heavy- and light-chain DNAs into a single mammalian expression vector, starting from DNAs encoding the desired variable and constant regions, which allows antibodies of different isotypes and specificity to be rapidly generated. As a proof of principle we have cloned, expressed and characterized functional recombinant tumor-associated antigen-specific chimeric IgE/κ and IgG1/κ, as well as recombinant grass pollen allergen Phl p 7 specific fully human IgE/λ and IgG4/λ antibodies. This method utilizing the antibody expression vectors, available at Addgene, has many applications, including the potential to support simultaneous processing of antibody panels, to facilitate mechanistic studies of antigen-antibody interactions and to conduct early evaluations of antibody functions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA002951
Anti-CSPG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA042785
Anti-CSPG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3701411 Anti-Human (κ-chain specific), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701417 Anti-Human (λ-chain specific), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F5512 Anti-Human IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F9512 Anti-Human IgG (Fc specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F1641 Anti-Human IgG (γ-chain specific), F(ab′)2 fragment–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F0132 Anti-Human IgG (γ-chain specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F6380 Anti-Human IgG (γ-chain specific)−FITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
F3512 Anti-Human IgG (whole molecule)−FITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
SAB1304953
ANTI-IGHG4 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1400136
Anti-IGHG4 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB4700351 Monoclonal Anti-IgE antibody produced in mouse, clone BE5, purified immunoglobulin, buffered aqueous solution
SAB4700352 Monoclonal Anti-IgE antibody produced in mouse, clone 4H10, purified immunoglobulin, buffered aqueous solution