EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Threonine-290 regulates nuclear translocation of the human pregnane X receptor through its phosphorylation/dephosphorylation by Ca2+/calmodulin-dependent protein kinase II and protein phosphatase 1.
PMID 25074870
Abstract
The human pregnane X receptor (hPXR) is recognized as a xenobiotic-sensing nuclear receptor that transcriptionally regulates the gene expression of drug-metabolizing enzymes and transporters. Our study elucidates the mechanism by which the localization of hPXR is regulated through threonine-290. A phosphomimetic mutation at threonine-290 (T290D) retained hPXR in the cytoplasm of HepG2, HuH6, and SW480 cells in vitro and the mouse liver in vivo even after treatment with rifampicin, and a phosphodeficient mutation (T290A) translocated from the cytoplasm to the nucleus as the wild-type hPXR. The amount of the unphosphorylated wild-type yellow fluorescent protein-hPXR fusion protein but not the T290A mutant increased on Phos-tag gels in response to stimulations with rifampicin and cyclin-dependent kinase 2 inhibitor roscovitine, and a marked increase was observed in the unphosphorylated levels of the T290A mutant in nontreated cells. The Ca(2+)/calmodulin-dependent protein kinase II (CaMKII) inhibitor KN93 [2-[N-(2-hydroxyethyl)]-N-(4-methoxybenzenesulfonyl)]amino-N-(4-chlorocinnamyl)-N-methylbenzylamine)] and transfection with anti-CaMKII small-interfering RNA (siRNA) enhanced the unphosphorylated levels of the wild-type protein. CaMKII directly phosphorylated the threonine-290 of hPXR, and the T290A mutant conferred resistance to CaMKII. The protein phosphatase (PP) inhibitor okadaic acid (100 nM) and transfection with anti-PP1 siRNA but not anti-PP2A siRNA led to reduced expression of CYP3A4 mRNA. After the rifampicin and roscovitine stimulations, PP1 was recruited to the wild-type hPXR but not the T290A mutant. These results suggest that phosphorylation at threonine-290 by CaMKII may impair the function of hPXR by repressing its translocation to the nucleus, and dephosphorylation by PP1 is necessary for the xenobiotic-dependent nuclear translocation of hPXR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406559
Anti-DNAJC7 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701904
Anti-dnajc7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA023015
Anti-DNAJC7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411368
Anti-DNAJC7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV46122
Anti-HIST1H1C antibody produced in rabbit, affinity isolated antibody
SAB2701559
Anti-HIST1H1C antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500408
Anti-Histone H1 (Acetyl-Lys25) antibody produced in rabbit, affinity isolated antibody
SAB2101093
Anti-HSP90AA1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2900334
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107614
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody
SAB2107615
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody
SAB4300541
Anti-HSP90AB1 (Ab-254) antibody produced in rabbit, affinity isolated antibody
SAB2701832
Anti-hsp90ab1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701806
Anti-hsp90ab1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV09016
Anti-HSP90AB1 antibody produced in rabbit, IgG fraction of antiserum
SAB2107344
Anti-HSP90AB1 antibody produced in rabbit, affinity isolated antibody
SAB4501462
Anti-HSP90B antibody produced in rabbit, affinity isolated antibody
SAB4501463
Anti-HSP90B antibody produced in rabbit, affinity isolated antibody
AV09018 Anti-HSPCB antibody produced in rabbit, IgG fraction of antiserum
SAB1305467
ANTI-HSPCB(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21241 Anti-HSPN antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1300598
Anti-PPP2R1A (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1401296
Anti-PPP2R1A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2103131
Anti-PPP2R1A, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1406288
Anti-PPP2R4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005695
Anti-PPP2R4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401298
Anti-PPP2R4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
61506
L-Threonine, certified reference material, TraceCERT®
C4H9NO3
PHR1242
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H9NO3
T1340000
L-Threonine, European Pharmacopoeia (EP) Reference Standard
C4H9NO3