EMAIL THIS PAGE TO A FRIEND
Proteome science
Automated quantitative multiplex immunofluorescence in situ imaging identifies phospho-S6 and phospho-PRAS40 as predictive protein biomarkers for prostate cancer lethality.
PMID 25075204
Abstract
We have witnessed significant progress in gene-based approaches to cancer prognostication, promising early intervention for high-risk patients and avoidance of overtreatment for low-risk patients. However, there has been less advancement in protein-based approaches, even though perturbed protein levels and post-translational modifications are more directly linked with phenotype. Most current, gene expression-based platforms require tissue lysis resulting in loss of structural and molecular information, and hence are blind to tumor heterogeneity and morphological features. Here we report an automated, integrated multiplex immunofluorescence in situ imaging approach that quantitatively measures protein biomarker levels and activity states in defined intact tissue regions where the biomarkers of interest exert their phenotype. Using this approach, we confirm that four previously reported prognostic markers, PTEN, SMAD4, CCND1 and SPP1, can predict lethal outcome of human prostate cancer. Furthermore, we show that two PI3K pathway-regulated protein activities, pS6 (RPS6-phosphoserines 235/236) and pPRAS40 (AKT1S1-phosphothreonine 246), correlate with prostate cancer lethal outcome as well (individual marker hazard ratios of 2.04 and 2.03, respectively). Finally, we incorporate these 2 markers into a novel 5-marker protein signature, SMAD4, CCND1, SPP1, pS6, and pPRAS40, which is highly predictive for prostate cancer-specific death. The ability to substitute PTEN with phospho-markers demonstrates the potential of quantitative protein activity state measurements on intact tissue. In summary, our approach can reproducibly and simultaneously quantify and assess multiple protein levels and functional activities on intact tissue specimens. We believe it is broadly applicable to not only cancer but other diseases, and propose that it should be well suited for prognostication at early stages of pathogenesis where key signaling protein levels and activities are perturbed.