EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Hypoxic conditions differentially regulate TAZ and YAP in cancer cells.
PMID 25078107
Abstract
The Hippo-YAP pathway is altered and implicated as an oncogenic signaling pathway in many human cancers. Hypoxia is an important microenvironmental factor that promotes tumorigenesis. However, the effects of hypoxia on the two most important Hippo-YAP effectors, YAP (Yes-associated protein) and TAZ (transcriptional co-activator with PDZ-binding motif), have not been reported. In this work, we demonstrated that TAZ was functionally involved in cell proliferation and/or migration in epithelial ovarian cancer (EOC) or human ovarian surface epithelial (HOSE) cells. Hypoxic conditions (1% O2 or hypoxia mimics) induced a reduction of YAP phosphorylation (S127) and total YAP expression in EOC cell lines OVCAR5 and SKOV3. However, these conditions up-regulated levels of S69 phosphorylated TAZ in EOC cells. The known TAZ kinases, Lats1 and Akt, were unlikely to be involved in up-regulated pTAZ by hypoxic conditions. Together, our data revealed new and differential regulating mechanisms of TAZ and YAP in cancer cells by hypoxia conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501013
Anti-Tafazzin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T4077 Anti-TAZ antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA039557
Anti-TAZ antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300644
Anti-TAZ antibody produced in rabbit, affinity isolated antibody
HPA007415
Anti-WWTR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32540 Anti-WWTR1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2102718
Anti-WWTR1 antibody produced in rabbit, affinity isolated antibody
EMU036491 MISSION® esiRNA, esiRNA targeting mouse Taz (esiRNA1)
EHU076701 MISSION® esiRNA, esiRNA human TAZ (esiRNA1)
EHU080321 MISSION® esiRNA, esiRNA human WWTR1 (esiRNA1)
EMU082641 MISSION® esiRNA, esiRNA targeting mouse Wwtr1 (esiRNA1)
WH0006901M12
Monoclonal Anti-TAZ antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution
SAB1404443
Monoclonal Anti-TAZ antibody produced in mouse, clone 1F11, purified immunoglobulin, buffered aqueous solution
SAB1409717 Monoclonal Anti-TAZ antibody produced in mouse, clone 3A12, purified immunoglobulin, buffered aqueous solution
SAB1409718
Monoclonal Anti-TAZ antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
SAB1409719 Monoclonal Anti-TAZ antibody produced in mouse, clone 3A11, purified immunoglobulin, buffered aqueous solution
SAB1409720 Monoclonal Anti-TAZ antibody produced in mouse, clone 1A4, purified immunoglobulin, buffered aqueous solution
SAB1409721
Monoclonal Anti-TAZ antibody produced in mouse, clone 3C10, purified immunoglobulin, buffered aqueous solution
SAB1409722
Monoclonal Anti-TAZ antibody produced in mouse, clone 1E5, purified immunoglobulin, buffered aqueous solution
AMAB90730
Monoclonal Anti-WWTR1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0371, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402558
Monoclonal Anti-WWTR1 antibody produced in mouse, clone 1F1, purified immunoglobulin, buffered aqueous solution