EMAIL THIS PAGE TO A FRIEND
Ultrastructural pathology
Ritonavir-induced hepatotoxicity and ultrastructural changes of hepatocytes.
PMID 25079492
Abstract
To investigate the effect of ritonavir on hepatocyte proliferation, we detected the change of cleaved caspase-3 expression level in the hepatocytes. Furthermore, the morphological and ultrastructural changes of hepatocytes derived from RTV-treated mice have been observed. The results showed that ritonavir can evidently inhibit hepatocyte proliferation and increase cleaved caspase-3 expression level. Under the electron microscope, chromatin margination, mitochondrial cristae disappearance, karyopyknosis and cytoplasmic vacuolization can be observed in the hepatocytes of mice treated with ritonavir. In conclusion, the mechanism of ritonavir's hepatotoxicity is that it induces apoptosis of hepatocytes via the caspase-cascade system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0491
Ritonavir, ≥98% (HPLC)
C37H48N6O5S2
Y0000714 Ritonavir, European Pharmacopoeia (EP) Reference Standard
C37H48N6O5S2