EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Effects of N source concentration and NH4(+)/NO3(-) ratio on phenylethanoid glycoside pattern in tissue cultures of Plantago lanceolata L.: a metabolomics driven full-factorial experiment with LC-ESI-MS(3.).
PMID 25081104
Abstract
Tissue cultures of a medicinal plant, Plantago lanceolata L. were screened for phenylethanoid glycosides (PGs) and other natural products (NPs) with LC-ESI-MS(3). The effects of N source concentration and NH4(+)/NO3(-) ratio were evaluated in a full-factorial (FF) experiment. N concentrations of 10, 20, 40 and 60mM, and NH4(+)/NO3(-) ratios of 0, 0.11, 0.20 and 0.33 (ratio of NH4(+) in total N source) were tested. Several peaks could be identified as PGs, of which, 16 could be putatively identified from the MS/MS/MS spectra. N source concentration and NH4(+)/NO3(-) ratio had significant effects on the metabolome, their effects on individual PGs were different despite these metabolites were of the same biosynthethic class. Chief PGs were plantamajoside and acteoside (verbascoside), their highest concentrations were 3.54±0.83% and 1.30±0.40% of dry weight, on media 10(0.33) and 40(0.33), respectively. NH4(+)/NO3(-) ratio and N source concentration effects were examined on a set of 89 NPs. For most NPs, high increases in abundance were observed compared to Murashige-Skoog medium. Abundances of 42 and 10 NPs were significantly influenced by the N source concentration and the NH4(+)/NO3(-) ratio, respectively. Optimal media for production of different NP clusters were 10(0), 10(0.11) and 40(0.33). Interaction was observed between NH4(+)/NO3(-) ratio and N source concentration for many NPs. It was shown in simulated experiments, that one-factor at a time (OFAT) experimental designs lead to sub-optimal media compositions for production of many NPs, and alternative experimental designs (e.g. FF) should be preferred when optimizing medium N source for optimal yield of NPs. If using OFAT, the N source concentration is to be optimized first, followed by NH4(+)/NO3(-) ratio, as this reduces the likeliness of suboptimal yield results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76514
2,4-D, certified reference material, TraceCERT®
C8H6Cl2O3
31518
2,4-D, PESTANAL®, analytical standard
C8H6Cl2O3
49083
2,4-D, analytical standard
C8H6Cl2O3
D7299
2,4-Dichlorophenoxyacetic acid, plant cell culture tested, ≥98%, crystalline
C8H6Cl2O3
D70724
2,4-Dichlorophenoxyacetic acid, 97%
C8H6Cl2O3
P9755
Pyridoxine hydrochloride, ≥98% (HPLC)
C8H11NO3 · HCl
P4722
Pyridoxine hydrochloride, meets USP testing specifications
C8H11NO3 · HCl
PHR1036
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C8H11NO3 · HCl
P4100000
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C8H11NO3 · HCl
1587001
Pyridoxine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C8H11NO3 · HCl
P6280
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C8H11NO3 · HCl
47862
Pyridoxine Hydrochloride (B6), analytical standard
C8H11NO3 · HCl
T4625
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
C12H17ClN4OS · HCl
T1270
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C12H17ClN4OS · HCl
51622
Thiamine hydrochloride, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C12H17ClN4OS · HCl
PHR1037
Thiamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C12H17ClN4OS · HCl
1656002
Thiamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C12H17ClN4OS · HCl
04573
Thiamine hydrochloride, tested according to Ph.Eur.
C12H17ClN4OS · HCl
T4562
Thiamine hydrochloride, meets USP testing specifications
C12H17ClN4OS · HCl
W332208
Thiamine hydrochloride, ≥98%, FCC, FG
C12H17ClN4OS · HCl
Y0000467
Thiamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C12H17ClN4OS · HCl
47858
Thiamine Hydrochloride (B1), analytical standard
C12H17ClN4OS · HCl