EMAIL THIS PAGE TO A FRIEND
Andrology
Mononuclear phagocytes rapidly clear apoptotic epithelial cells in the proximal epididymis.
PMID 25082073
Abstract
We have shown previously that a network of mononuclear phagocytes (MPs) expressing macrophage and dendritic cell markers such as CD11c, F4/80 and CX3CR1, lines the base of the epididymal tubule. However, in the initial segment (IS) and only in that particular segment, epididymal MPs establish extremely close interactions with the epithelium by projecting slender dendrites between most epithelial cells. We undertook the present study to determine how epididymal phagocytes respond to the transient wave of apoptosis initiated by unilateral efferent duct ligation (EDL) in the epididymal epithelium. We show profound morphological and phenotypical changes restricted to the MPs populating the proximal epididymis following EDL. Within 48 h, a large subset of IS epithelial cells had entered an apoptotic state, visualized by the terminal deoxynucleotidyl transferase dUTP nick end labelling (TUNEL) assay and CD11c(+) and CX3CR1(+) MPs readily engulfed TUNEL-positive cells and other debris. Despite the high levels of apoptosis and the rapid clearance of apoptotic cells occurring after EDL, the epithelium preserved its overall architecture and maintained tight junctions of the blood-epididymis barrier (BEB). The discovery of a functional population of MPs in the epididymal epithelium responsible for maintaining the integrity of the BEB raises further questions regarding the role of these cells in clearing defective epithelial cells in the steady-state epididymis, as well as pathogens and abnormal spermatozoa in the lumen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406503
Anti-T antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1412153
ANTI-T antibody produced in mouse, clone 5C6, purified immunoglobulin, buffered aqueous solution
SAB1412159
ANTI-T antibody produced in mouse, clone 5C10, purified immunoglobulin, buffered aqueous solution
SAB1412193
ANTI-T antibody produced in mouse, clone 5C8, purified immunoglobulin, buffered aqueous solution
SAB1412313
ANTI-T antibody produced in mouse, clone 5E11, purified immunoglobulin, buffered aqueous solution
SAB1412314
ANTI-T antibody produced in mouse, clone 6D3, purified immunoglobulin, buffered aqueous solution
SAB1412315
ANTI-T antibody produced in mouse, clone 5E12, purified immunoglobulin, buffered aqueous solution
SAB1412316
ANTI-T antibody produced in mouse, clone 2A12, purified immunoglobulin, buffered aqueous solution
SAB1412317
ANTI-T antibody produced in mouse, clone 1H1, purified immunoglobulin, buffered aqueous solution
HPA003322
Anti-T antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104044
Anti-T antibody produced in rabbit, affinity isolated antibody
WH0006862M2
Monoclonal Anti-T antibody produced in mouse, clone 5C5, purified immunoglobulin, buffered aqueous solution
SAB1404434
Monoclonal Anti-T antibody produced in mouse, clone 5H8, purified immunoglobulin, buffered aqueous solution