EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
Phenolic composition and antioxidant capacity of pomaces from four grape varieties (Vitis vinifera L.).
PMID 25082193
Abstract
Phenolic compounds are widely distributed secondary metabolites in plants usually conferring them with unique taste, flavour and health-promoting properties. In fruits of Vitis vinifera L., phenolic composition is highly dependent on grape variety. Differential extraction of these compounds from grapes during winemaking is critically associated with wine quality. By-products of winemaking, such as grape pomace, can contain significant amounts of polyphenols. However, information concerning the varietal effect on wine grape pomace is scarce. In this study, pomaces from Sauvignon Blanc (SB), Chardonnay (CH), Cabernet Sauvignon (CS) and Carménère (CA) grape varieties were characterized spectroscopically and by HPLC-DAD analysis. White grape pomaces (SB and CH) presented higher antioxidant capacities and higher contents of total phenols and total proanthocyanidins compared with red grape pomaces (CS and CA), whereas the latter showed much higher anthocyanin levels and colour intensities. Concentrations of monomeric proanthocyanidins and low-molecular-weight phenols in the four grape pomace varieties were significantly different. Grape pomaces from four varieties showed high but diverse contents of polyphenols and antioxidant capacities. Thus grape pomaces represent an important potential source of polyphenols, which could be useful for nutritional and/or pharmacological purposes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1251
(+)-Catechin hydrate, ≥98% (HPLC), powder
C15H14O6 · xH2O
E3893
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
C22H18O10
37580
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
C7H6O4
08992
3,4-Dihydroxybenzoic acid, analytical standard
C7H6O4
58958
Ethyl acetate, analytical standard
C4H8O2
17023
Ethyl acetate, tested according to Ph.Eur.
C4H8O2
PHR1481
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
C4H8O2
G7384
Gallic acid, 97.5-102.5% (titration)
C7H6O5
91215
Gallic acid, certified reference material, TraceCERT®
C7H6O5
96353
Kaempferol, analytical standard
C15H10O6
00550580
Kaempferol, primary pharmaceutical reference standard
C15H10O6
60010
Kaempferol, ≥97.0% (HPLC)
C15H10O6
K0133
Kaempferol, ≥90% (HPLC), powder
C15H10O6
77634
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
C8H8O3
72576
Myricetin, analytical standard
C15H10O8
M6760
Myricetin, ≥96.0%, crystalline
C15H10O8
70050
Myricetin, ≥96.0% (HPLC)
C15H10O8
03930590
Protocatechuic acid, primary pharmaceutical reference standard
C7H6O4
PHR1488
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
C15H10O7 · xH2O
Q4951
Quercetin, ≥95% (HPLC), solid
C15H10O7
1592409
Quercetin, United States Pharmacopeia (USP) Reference Standard
C15H10O7 · 2H2O
94770
Vanillic acid, purum, ≥97.0% (HPLC)
C8H8O4
H36001
Vanillic acid, 97%
C8H8O4
W398802
Vanillic acid, ≥97%, FG
C8H8O4
68654
Vanillic acid, certified reference material, TraceCERT®
C8H8O4
V2375
Vanillin
C8H8O3
30304
Vanillin, TraceCERT®, certified reference material
C8H8O3
94752
Vanillin, tested according to Ph.Eur.
C8H8O3
PHR1245
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
V1104
Vanillin, ReagentPlus®, 99%
C8H8O3