EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
Light-induced termination of spiral wave arrhythmias by optogenetic engineering of atrial cardiomyocytes.
PMID 25082848
Abstract
Atrial fibrillation (AF) is the most common cardiac arrhythmia and often involves reentrant electrical activation (e.g. spiral waves). Drug therapy for AF can have serious side effects including proarrhythmia, while electrical shock therapy is associated with discomfort and tissue damage. Hypothetically, forced expression and subsequent activation of light-gated cation channels in cardiomyocytes might deliver a depolarizing force sufficient for defibrillation, thereby circumventing the aforementioned drawbacks. We therefore investigated the feasibility of light-induced spiral wave termination through cardiac optogenetics. Neonatal rat atrial cardiomyocyte monolayers were transduced with lentiviral vectors encoding light-activated Ca(2+)-translocating channelrhodopsin (CatCh; LV.CatCh∼eYFP↑) or eYFP (LV.eYFP↑) as control, and burst-paced to induce spiral waves rotating around functional cores. Effects of CatCh activation on reentry were investigated by optical and multi-electrode array (MEA) mapping. Western blot analyses and immunocytology confirmed transgene expression. Brief blue light pulses (10 ms/470 nm) triggered action potentials only in LV.CatCh∼eYFP↑-transduced cultures, confirming functional CatCh-mediated current. Prolonged light pulses (500 ms) resulted in reentry termination in 100% of LV.CatCh∼eYFP↑-transduced cultures (n = 31) vs. 0% of LV.eYFP↑-transduced cultures (n = 11). Here, CatCh activation caused uniform depolarization, thereby decreasing overall excitability (MEA peak-to-peak amplitude decreased 251.3 ± 217.1 vs. 9.2 ± 9.5 μV in controls). Consequently, functional coresize increased and phase singularities (PSs) drifted, leading to reentry termination by PS-PS or PS-boundary collisions. This study shows that spiral waves in atrial cardiomyocyte monolayers can be terminated effectively by a light-induced depolarizing current, produced by the arrhythmogenic substrate itself, upon optogenetic engineering. These results provide proof-of-concept for shockless defibrillation.