EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Sexually dimorphic RB inactivation underlies mesenchymal glioblastoma prevalence in males.
PMID 25083989
Abstract
The prevalence of brain tumors in males is common but unexplained. While sex differences in disease are typically mediated through acute sex hormone actions, sex-specific differences in brain tumor rates are comparable at all ages, suggesting that factors other than sex hormones underlie this discrepancy. We found that mesenchymal glioblastoma (Mes-GBM) affects more males as the result of cell-intrinsic sexual dimorphism in astrocyte transformation. We used astrocytes from neurofibromin-deficient (Nf1(-/-)) mice expressing a dominant-negative form of the tumor suppressor p53 (DNp53) and treated them with EGF as a Mes-GBM model. Male Mes-GBM astrocytes exhibited greater growth and colony formation compared with female Mes-GBM astrocytes. Moreover, male Mes-GBM astrocytes underwent greater tumorigenesis in vivo, regardless of recipient mouse sex. Male Mes-GBM astrocytes exhibited greater inactivation of the tumor suppressor RB, higher proliferation rates, and greater induction of a clonogenic, stem-like cell population compared with female Mes-GBM astrocytes. Furthermore, complete inactivation of RB and p53 in Mes-GBM astrocytes resulted in equivalent male and female tumorigenic transformation, indicating that intrinsic differences in RB activation are responsible for the predominance of tumorigenic transformation in male astrocytes. Together, these results indicate that cell-intrinsic sex differences in RB regulation and stem-like cell function may underlie the predominance of GBM in males.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4501977
Anti-NF1 antibody produced in rabbit, affinity isolated antibody
SAB1305397 ANTI-PHOSPHO-RB(S249) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305092 ANTI-PHOSPHO-RB(S608) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305398 ANTI-PHOSPHO-RB(S612) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305093 ANTI-PHOSPHO-RB(S780) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305094 ANTI-PHOSPHO-RB(S788) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305095
ANTI-PHOSPHO-RB(S811) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300109
Anti-phospho-RB1 (pSer780) antibody produced in rabbit, affinity isolated antibody
SAB4300107
Anti-phospho-RB1 (pSer795) antibody produced in rabbit, affinity isolated antibody
SAB4300108
Anti-phospho-RB1 (pSer807) antibody produced in rabbit, affinity isolated antibody
SAB4300387
Anti-RB1 (Ab-780) antibody produced in rabbit, affinity isolated antibody
SAB4300385
Anti-RB1 (Ab-795) antibody produced in rabbit, affinity isolated antibody
SAB4300386
Anti-RB1 (Ab-807) antibody produced in rabbit, affinity isolated antibody
AV33212
Anti-RB1 antibody produced in rabbit, affinity isolated antibody
WH0000836M2 Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder